Skip to Content
Merck
All Photos(2)

Key Documents

06-1385

Sigma-Aldrich

Anti-PDX1 (goat) Antibody

serum, from goat

Synonym(s):

pancreatic and duodenal homeobox 1, pancreatic-duodenal homeobox factor 1, somatostatin transcription factor 1, insulin promoter factor 1, homeodomain transcription factor, Insulin promoter factor 1, Somatostatin-transactivating factor 1, Insulin upstrea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
polyclonal
application:
IHC
WB
species reactivity:
human, mouse
technique(s):
immunohistochemistry: suitable
western blot: suitable
citations:
4

biological source

goat

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Pdx1(18609)

General description

Transcription factor PDX1, also known as pancreatic and duodenal homeobox 1, activates expression of the insulin and somatostatin genes. This protein is a key regulator of islet peptide hormone expression and also plays an essential role in pancreatic development. It has been shown to interact with the basic helix-loop-helix domains of TCF3(E47) and NEUROD1 and with HMG-I(Y). This protein also interacts with the methyltransferase SETD7. Mutations in this gene may be involved in several disorders of the pancreas or in diabetes mellitus.

Immunogen

Recombinant protein corresponding to the N-terminus of mouse PDX1.

Application

Anti-PDX1 (goat) Antibody is a Goat Polyclonal Antibody for detection of PDX1 also known as pancreatic & duodenal homeobox 1, somatostatin transcription factor 1, Insulin promoter factor 1 & has been validated in WB & IHC.
Immunohistochemistry Analysis: 1:400 dilution from a represenative lot detected PDX1 in normal human pancreas tissue.

Quality

Evaluated by Western Blot in mouse pancreatic β T-cell lysate.

Western Blot Analysis: 1:1,000 dilution of this antibody detected PDX1 on 10 µg of mouse pancreatic β T-cell lysate.

Target description

~ 39 kDa

Physical form

Unpurified goat polyclonal serum containing 0.05% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Romina J Bevacqua et al.
Nature communications, 12(1), 2397-2397 (2021-04-25)
Gene targeting studies in primary human islets could advance our understanding of mechanisms driving diabetes pathogenesis. Here, we demonstrate successful genome editing in primary human islets using clustered regularly interspaced short palindromic repeats (CRISPR) and CRISPR-associated protein 9 (Cas9). CRISPR-based
Signaling pathways for sphingosylphosphorylcholine-mediated mitogenesis in Swiss 3T3 fibroblasts.
N N Desai,R O Carlson,M E Mattie,A Olivera,N E Buckley,T Seki,G Brooker,S Spiegel
The Journal of cell biology null
Giannicola Genovese et al.
Nature, 542(7641), 362-366 (2017-02-09)
Malignant neoplasms evolve in response to changes in oncogenic signalling. Cancer cell plasticity in response to evolutionary pressures is fundamental to tumour progression and the development of therapeutic resistance. Here we determine the molecular and cellular mechanisms of cancer cell
Pauline Jeannot et al.
Oncotarget, 6(34), 35880-35892 (2015-09-30)
p27Kip1 (p27) is a negative regulator of proliferation and a tumor suppressor via the inhibition of cyclin-CDK activity in the nucleus. p27 is also involved in the regulation of other cellular processes, including transcription by acting as a transcriptional co-repressor.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service