Skip to Content
Merck
All Photos(1)

Key Documents

V-025

Supelco

Vitamin D3 solution

1 mg/mL in ethanol, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

Vitamin D3 solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H44O
CAS Number:
Molecular Weight:
384.64
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

(Snap-N-Shoot®)
(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1 mg/mL in ethanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing
clinical testing

format

single component solution

storage temp.

−70°C

SMILES string

O[C@H]1CCC(=C)\C(=C/C=C2/[C@H]3[C@@]([C@H](CC3)[C@@H](CCCC(C)C)C)(CCC/2)C)\C1

InChI

1S/C27H44O/c1-19(2)8-6-9-21(4)25-15-16-26-22(10-7-17-27(25,26)5)12-13-23-18-24(28)14-11-20(23)3/h12-13,19,21,24-26,28H,3,6-11,14-18H2,1-2,4-5H3/b22-12+,23-13-/t21-,24+,25-,26+,27-/m1/s1

InChI key

QYSXJUFSXHHAJI-YRZJJWOYSA-N

Gene Information

human ... VDR(7421)

General description

Vitamin D3 or cholecalciferol is a form of vitamin D produced naturally by the skin when exposed to ultraviolet light. Vitamin D3 is also manufactured industrially to fortify foods or for use in vitamin supplements.

Application

This certified solution standard is suitable for use as a Snap-N-Spike® starting material in the preparation of calibrators or controls for clinical applications or Snap-N-Shoot® in quality control testing of vitamin supplements, fortified foods or pharmaceutical preparations.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

57.2 °F

Flash Point(C)

14.0 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuan-Hua Chen et al.
Toxicological sciences : an official journal of the Society of Toxicology, 145(1), 90-97 (2015-02-13)
Several reports demonstrated that maternal lipopolysaccharide (LPS) exposure at middle gestational stage caused neural tube defects (NTDs). This study investigated the effects of supplementation with vitamin D3 (VitD3) during pregnancy on LPS-induced NTDs. Pregnant mice except controls were ip injected
Stefan Pilz et al.
Hypertension (Dallas, Tex. : 1979), 65(6), 1195-1201 (2015-03-25)
Vitamin D deficiency is a risk factor for arterial hypertension, but randomized controlled trials showed mixed effects of vitamin D supplementation on blood pressure (BP). We aimed to evaluate whether vitamin D supplementation affects 24-hour systolic ambulatory BP monitoring values
Robin M Daly et al.
The American journal of clinical nutrition, 99(4), 899-910 (2014-01-31)
Physical inactivity, inadequate dietary protein, and low-grade systemic inflammation contribute to age-related muscle loss, impaired function, and disability. We assessed the effects of progressive resistance training (PRT) combined with a protein-enriched diet facilitated through lean red meat on lean tissue
Anders Burild et al.
Steroids, 98, 72-79 (2015-03-01)
Information regarding the endogenous storages of vitamin D3 after cutaneous vitamin D synthesis compared to oral vitamin D3 supplementation is sparse. Furthermore it is not known whether vitamin D3 can be stored for later use during periods of shortages of
Negar Tabatabaei et al.
The Journal of nutrition, 144(12), 1994-2001 (2014-10-25)
Whether there is a dose-dependent effect of maternal dietary cholecalciferol during pregnancy on maternal glucose tolerance is unknown. In addition, circulating osteocalcin is increased by 1,25-dihydroxyvitamin D [1,25(OH)2D] and may improve glucose homeostasis. This study was designed to test whether

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service