Skip to Content
Merck
All Photos(1)

Documents

P-084

Supelco

(±)-Pentazocine-13C3 hydrochloride

100 μg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

(±)-Pentazocine-13C3 hydrochloride solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C3C16H27NO · HCl
Molecular Weight:
324.86
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule A (Switzerland); psicótropo (Spain); Decreto Lei 15/93: Tabela IIC (Portugal)

concentration

100 μg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

O=C(C1=C2C(NC(CCOCCC(O)=O)=O)=CC=C1)N([C@H]3CCC(NC3=O)=O)C2=O.O=C(CCOCCC(O)=O)N[C@@H](C(C)(C)C)C(N4C[C@H](O)C[C@H]4C(NCC5=CC=C(C6=C(C)N=CS6)C=C5)=O)=O

InChI

1S/C19H27NO.ClH/c1-13(2)7-9-20-10-8-19(4)14(3)18(20)11-15-5-6-16(21)12-17(15)19;/h5-7,12,14,18,21H,8-11H2,1-4H3;1H/i1+1,2+1,13+1;

InChI key

OQGYMIIFOSJQSF-NASIXXEGSA-N

General description

Pentazocine is a synthetic opioid analgesic sold under trade names Fortral, Talwin, or Talacen and used to treat moderate to moderately severe pain. Suitable use of this certified Snap-N-Spike® solution and internal standard includes quantitation of pentazocine levels and isotope dilution methods by GC/MS or LC/MS in applications for clinical toxicology, urine drug testing, forensic analysis or pain prescription monitoring.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service