Skip to Content
Merck
All Photos(1)

Key Documents

H-003

Supelco

Hydrocodone solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H21NO3
CAS Number:
Molecular Weight:
299.36
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule A (Switzerland); estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal)

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

N1([C@H]2[C@H]3[C@@]4([C@@H](Oc5c4c(ccc5OC)C2)C(=O)CC3)CC1)C.O[C@H]([C@@H](O)C(=O)O)C(=O)O.O

InChI

1S/C18H21NO3.C4H6O6.H2O/c1-19-8-7-18-11-4-5-13(20)17(18)22-16-14(21-2)6-3-10(15(16)18)9-12(11)19;5-1(3(7)8)2(6)4(9)10;/h3,6,11-12,17H,4-5,7-9H2,1-2H3;1-2,5-6H,(H,7,8)(H,9,10);1H2/t11-,12+,17-,18-;1-,2-;/m01./s1

InChI key

ZICRJLQTAUVSDU-NSJZFRMTSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hydrocodone is a semi-synthetic opioid derived from codeine or thebaine, two naturally-occurring opiates. The drug, also known as dihydrocodeinone, is used as an analgesic to treat pain and as an antitussive for coughing or related conditions. This Certified Spiking Solution® is suitable as a starting material for calibrators or controls in GC/MS or LC/MS testing applications from pharmaceutical research and pain prescription monitoring to clinical toxicology and forensic analysis.

Application


  • Hydrocodone Bitartrate ER: A Review in Chronic Pain.: This comprehensive review evaluates the extended-release formulation of hydrocodone, focusing on its pharmacological properties, therapeutic efficacy, and clinical applications in managing chronic pain, which underscores its significance in long-term pain management strategies (Dhillon S, 2016).

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

FDA panel recommends tighter rules for prescribing opioids.
Michael McCarthy
BMJ (Clinical research ed.), 346, f1012-f1012 (2013-02-16)
Kathryn P Anastassopoulos et al.
Journal of opioid management, 9(2), 97-109 (2013-05-28)
Pain is a prevalent condition that often involves a neuropathic component. Hydrocodone is one of the most widely used opioids for pain but is often associated with side effects (SEs). This study sought to characterize the experience of patients taking
Elizabeth A Micks et al.
Obstetrics and gynecology, 120(5), 1060-1069 (2012-10-24)
Although hydrocodone-acetaminophen is commonly used for pain control in first-trimester abortion, the efficacy of oral opioids for decreasing pain has not been established. Our objective was to estimate the effect of hydrocodone-acetaminophen on patient pain perception during first-trimester surgical abortion.
How well can urine hydrocodone measurements discriminate between different hydrocodone prescription dosage rates?
Laura J McCloskey et al.
Clinica chimica acta; international journal of clinical chemistry, 419, 119-121 (2013-02-23)
William Edward Ackerman
The Journal of the Arkansas Medical Society, 109(5), 90-93 (2012-10-16)
Hydrocodone is frequently prescribed for moderate to severe pain. Nicotine has analgesic properties and it is hypothesized that cigarette smoking might decrease the dosage of hydrocodone needed for the relief of chronic pain. This would be a topic of importance

Articles

Although both biphenyl and phenyl-hexyl phases can resolve these compounds, the former exhibits excellent peak shape and substantially less silanol-derived ion exchange activity.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service