Skip to Content
Merck
All Photos(1)

Key Documents

H-038

Supelco

Heroin solution

1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H23NO5
CAS Number:
Molecular Weight:
369.41
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule D (Switzerland); estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal)

concentration

1.0 mg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

CN1CC[C@]23[C@H]4Oc5c(OC(C)=O)ccc(C[C@@H]1[C@@H]2C=C[C@@H]4OC(C)=O)c35

InChI

1S/C21H23NO5/c1-11(23)25-16-6-4-13-10-15-14-5-7-17(26-12(2)24)20-21(14,8-9-22(15)3)18(13)19(16)27-20/h4-7,14-15,17,20H,8-10H2,1-3H3/t14-,15+,17-,20-,21-/m0/s1

InChI key

GVGLGOZIDCSQPN-PVHGPHFFSA-N

General description

Heroin, or diacetylmorphine, is an opiate analgesic most commonly used as an illcit recreational drug. Street heroin is considered to be one of the most harmful recreational drugs due to the use of adulterants and the risk of contracting disease from intravenous use. This drug reference standard is suitable for use in GC/MS or LC/MS heroin testing methods for applications in clinical toxicology, pain prescription monitoring, forensic analysis, and urine drug testing.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

De-Xiang Li et al.
The American journal of Chinese medicine, 41(1), 43-57 (2013-01-23)
While exercise has been shown to reduce the negative effects of substance withdrawal symptoms, no research has investigated if Tai Chi, a traditional Chinese exercise, has similar effects. Here, we observed the physiological effects of Tai Chi on protracted abstinence
Albert Sánchez-Niubò et al.
BMC medical research methodology, 13, 4-4 (2013-01-16)
Existing incidence estimates of heroin use are usually based on one information source. This study aims to incorporate more sources to estimate heroin use incidence trends in Spain between 1971 and 2005. A multi-state model was constructed, whereby the initial
Ruixue Yin et al.
International journal of medical sciences, 10(3), 299-306 (2013-02-21)
To investigate whether microvascular damage is involved in the pathogenesis of heroin induced spongiform leukoencephalopathy (HSLE). The brain tissues were collected from 4 HSLE patients and 5 controls and then fixed in 4% paraformaldehyde. The frontal lobe, corpus callosum and
Hannah Skuse et al.
Emergency nurse : the journal of the RCN Accident and Emergency Nursing Association, 20(9), 14-19 (2013-03-23)
This article discusses the administration of intranasal diamorphine as an analgesia-in paediatric trauma care. The authors outline evidence for its use and discuss a recent multicentre safety study of an intranasal diamorphine product. In addition, they advise emergency department nurses
Marc Walter et al.
Journal of clinical psychopharmacology, 33(2), 193-198 (2013-02-21)
Heroin dependence is associated with a stressful environment and with dysfunction of the hypothalamic-pituitary-adrenal (HPA) axis. The present study examined the acute effects of intravenous heroin versus placebo on the HPA axis response in heroin-dependent patients. Twenty-eight heroin-dependent patients in

Protocols

Analytical standards for drug analysis including heroin, cocaine, amphetamine, and methylenedioxymethamphetamine.

Analytical standards for drug analysis including heroin, cocaine, amphetamine, and methylenedioxymethamphetamine.

Analytical standards for drug analysis including heroin, cocaine, amphetamine, and methylenedioxymethamphetamine.

Analytical standards for drug analysis including heroin, cocaine, amphetamine, and methylenedioxymethamphetamine.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service