Skip to Content
Merck
All Photos(1)

Key Documents

D-035

Supelco

Diltiazem hydrochloride solution

1.0 mg/mL in acetonitrile (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H27ClN2O4S
CAS Number:
Molecular Weight:
450.98
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in acetonitrile (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

2-8°C

SMILES string

Cl.COc1ccc(cc1)[C@@H]2Sc3ccccc3N(CCN(C)C)C(=O)[C@@H]2OC(C)=O

InChI

1S/C22H26N2O4S.ClH/c1-15(25)28-20-21(16-9-11-17(27-4)12-10-16)29-19-8-6-5-7-18(19)24(22(20)26)14-13-23(2)3;/h5-12,20-21H,13-14H2,1-4H3;1H/t20-,21+;/m1./s1

InChI key

HDRXZJPWHTXQRI-BHDTVMLSSA-N

General description

A Certified Spiking Solution® produced for use in clinical toxicology, forensic analysis, or pharmaceutical research applications by GC/Ms or LC/MS. Diltiazem is a pharmaceutical drug in the calcium channel blocker class. Diltiazem is prescribed for the treatment of hypertension and heart conditions such as angina pectoris and some types of arrhythmia.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jasper Dingemanse et al.
Clinical drug investigation, 33(3), 207-213 (2013-02-06)
The cytochrome P450 (CYP) enzyme, CYP3A4, metabolizes ACT-178882, a new direct renin inhibitor. This study investigated the effect of diltiazem, a moderate inhibitor of CYP3A4, on the single-dose pharmacokinetics of ACT-178882 in healthy subjects. In this open-label, two-way crossover, drug-drug
Mustafa Kayan et al.
The Journal of membrane biology, 245(12), 833-840 (2012-08-21)
Non-ionic contrast media (CM) can induce tissue kidney injury via activation of phagocytosis and oxidative stress, although the mechanisms of injury via neutrophils are not clear. We investigated the effects of CM on oxidative stress and Ca²⁺ concentrations in serum
T Zheng et al.
Die Pharmazie, 68(4), 257-260 (2013-05-25)
To assess the possibility of using CYP2D6 10 +/- CYP3A5*3 as biomarkers to predict the pharmacokinetics of diltiazem and its two metabolites among healthy Chinese subjects. METHODS 41 healthy Chinese were genotyped for CYP3A5 3 and CYP2D6 10, and then
Ronald W Day
Congenital heart disease, 8(1), 71-77 (2012-08-16)
Right heart catheterization is performed in patients with pulmonary arterial hypertension to determine the severity of disease and their pulmonary vascular reactivity. This study sought to determine whether the acute pulmonary vasodilatory effects of diltiazem and oxygen with nitric oxide
[Optimum heart rate - the current goal of cardiovascular therapy. Position statement of the Polish Cardiac Society Working Group on Cardiovascular Drug Therapy].
Jarosław D Kasprzak et al.
Kardiologia polska, 70(10), 1081-1094 (2012-10-20)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service