Skip to Content
Merck
All Photos(1)

Key Documents

860518P

Avanti

C18 Ceramide (d18:1/18:0)

Avanti Research - A Croda Brand

Synonym(s):

N-stearoyl-D-erythro-sphingosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H71NO3
CAS Number:
Molecular Weight:
565.95
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 10 mg (860518P-10mg)
pkg of 1 × 5 mg (860518P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

OC[C@]([H])(NC(CCCCCCCCCCCCCCCCC)=O)[C@]([H])(O)/C=C/CCCCCCCCCCCCC

InChI

1S/C36H71NO3/c1-3-5-7-9-11-13-15-17-18-20-22-24-26-28-30-32-36(40)37-34(33-38)35(39)31-29-27-25-23-21-19-16-14-12-10-8-6-4-2/h29,31,34-35,38-39H,3-28,30,32-33H2,1-2H3,(H,37,40)/b31-29+/t34-,35+/m0/s1

InChI key

VODZWWMEJITOND-NXCSZAMKSA-N

General description

C18 Ceramide (d18:1/18:0) synthesis is catalyzed by ceramide synthase (CERS) 1 and 5. It is a component of cellular membrane and has low gel forming ability compared to unsaturated ceramide.

Application

C18 Ceramide (d18:1/18:0) has been used:
  • to induce apoptosis in Manduca embryonic cell line (MRRL-CH1)
  • as a standard to quantify ceramide from liver tissue using liquid chromatography-mass spectrometry
  • to inhibit ceramidase in human embryonic kidney cells (HEK29) with β-glucocerebrosidase (GCase) mutation

Biochem/physiol Actions

C18 Ceramide based derivatives are present in low levels during neurodegeneration. It favors endoplasmic reticulum based stress and autophagy in glioma cells. Low levels of C18-ceramide are implicated in human head and neck squamous cell carcinoma. C18 ceramide may inhibit tumor progression.

Packaging

5 mL Amber Glass Screw Cap Vial (860518P-10mg)
5 mL Amber Glass Screw Cap Vial (860518P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Overexpression of ceramide synthase 1 increases C18-ceramide and leads to lethal autophagy in human glioma
Wang Z, et al.
Testing, 8(61), 104022-104022 (2017)
Overexpression of ceramide synthase 1 increases C18-ceramide and leads to lethal autophagy in human glioma
Koybasi S, et al.
The Journal of biological chemistry, 279(43), 44311-44319 (2004)
D T U Abeytunga et al.
Journal of lipid research, 45(7), 1221-1231 (2004-04-23)
NMR and electrospray ionization tandem mass spectrometry were used to show for the first time the presence of sphingomyelins in extracts of the tobacco hornworm Manduca sexta (Lepidoptera). The sphingosine in the ceramide was identified as tetradecasphing-4-enine, and the fatty
Acid ceramidase inhibition ameliorates alpha-synuclein accumulation upon loss of GBA1 function
Kim MJ, et al.
Human Molecular Genetics, 27(11), 1972-1988 (2018)
Presence of unsaturated sphingomyelins and changes in their composition during the life cycle of the moth Manduca sexta
Abeytunga DTU, et al.
Journal of Lipid Research, 45(7), 1221-1231 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service