860512P
Avanti
C12 Ceramide (d18:1/12:0)
Avanti Research™ - A Croda Brand
Synonym(s):
N-lauroyl-D-erythro-sphingosine
About This Item
Recommended Products
form
powder
packaging
pkg of 1 × 10 mg (860512P-10mg)
pkg of 1 × 25 mg (860512P-25mg)
pkg of 1 × 5 mg (860512P-5mg)
manufacturer/tradename
Avanti Research™ - A Croda Brand
lipid type
sphingolipids
shipped in
dry ice
storage temp.
−20°C
SMILES string
OC[C@]([H])(NC(CCCCCCCCCCC)=O)[C@]([H])(O)/C=C/CCCCCCCCCCCCC
InChI
1S/C30H59NO3/c1-3-5-7-9-11-13-14-15-16-18-19-21-23-25-29(33)28(27-32)31-30(34)26-24-22-20-17-12-10-8-6-4-2/h23,25,28-29,32-33H,3-22,24,26-27H2,1-2H3,(H,31,34)/b25-23+/t28-,29+/m0/s1
InChI key
HXFPPRPLRSPNIB-VARSQMIESA-N
General description
Application
- as a lipid standard in the optimization of the liquid chromatography-mass spectrometry (LC-MS) method conditions
- as an internal standard for lipid extraction in U87MG (human glioma cell line)
- as an internal standard for the quantification of ceramide using liquid chromatography-mass spectrometry (LC-MS) in rat tissues
Biochem/physiol Actions
Packaging
Legal Information
also commonly purchased with this product
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service