Skip to Content
Merck
All Photos(2)

Key Documents

Z692549

Scienceware® Secador® auto-desiccator cabinet

model 4.0, vertical profile, amber, AC/DC input 230 V AC, CE compliant

Synonym(s):

Bell Jar, Desiccator, Desiccator cabinet

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104510
NACRES:
NB.43

AC/DC input

, universal plug set (includes Europlug and GB plug)

material

polycarbonate

feature

CE compliant

packaging

pack of 1 ea

manufacturer/tradename

Bel-Art F42074-1228

H × W × D

51.8 cm × 34.1 cm × 41.4 cm

door opening H × W

41.7 cm × 22.5 cm

color

amber

Looking for similar products? Visit Product Comparison Guide

General description

Approved by both CSA and UL

Protecting valuable humidity sensitive items such as reagents or pipettes is easy and totally automatic with Secador Auto-Desiccator Cabinets. Three different size models are available to best match individual storage needs, and can be stacked together to save space.

The 2.0, 3.0 and 4.0 models have internal volumes of 1.1 cu.ft., 1.5 cu. ft., and 1.9 cu.ft. respectively. The cabinets are molded of rugged polycarbonate resistant to staining, crazing and chemical attack.

A patented electronic dehumidification system is designed to reduce the relative humidity inside the cabinet from 75 to 25% and circulate dry air automatically, freeing users form monitoring or changing chemical desiccant.

All contents are easily viewed through a large clear door that seals with positive latches and can be securely closed with tamper evident seals. A hygrometer in the door allows for easy monitoring of relative humidity.

Includes: Perforated, removable shelves.

Other Notes

Supplied with: interchangeable power plugs

Legal Information

Scienceware is a registered trademark of SP Scienceware, a division of SP
Secador is a registered trademark of SP Scienceware, a division of SP

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service