Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Scienceware
®
Secador
®
auto-desiccator cabinet
Products
Product Category
desiccators (12)
Brand
Sigma-Aldrich (1)
Color
Manufacturer
Package Size
Filter Products
Showing 1-12 of 12
Compare
Product Number
Description
Pricing
Z692506
Scienceware
®
Secador
®
auto-desiccator cabinet,
model 3.0, vertical profile, clear, AC/DC input 230 V AC, CE compliant
Expand
View Pricing
Z553794
Scienceware
®
Secador
®
auto-desiccator cabinet,
model 3.0, vertical profile, clear, AC/DC input 120 V AC
Expand
View Pricing
Z692549
Scienceware
®
Secador
®
auto-desiccator cabinet,
model 4.0, vertical profile, amber, AC/DC input 230 V AC, CE compliant
Expand
View Pricing
Z692492
Scienceware
®
Secador
®
auto-desiccator cabinet,
model 2.0, square profile, clear, AC/DC input 230 V AC, CE compliant
Expand
View Pricing
Z692530
Scienceware
®
Secador
®
auto-desiccator cabinet,
model 3.0, vertical profile, amber, AC/DC input 230 V AC, CE compliant
Expand
View Pricing
Z692514
Scienceware
®
Secador
®
auto-desiccator cabinet,
model 4.0, vertical profile, clear, AC/DC input 230 V AC, CE compliant
Expand
View Pricing
Z692522
Scienceware
®
Secador
®
auto-desiccator cabinet,
model 2.0, square profile, amber, AC/DC input 230 V AC, CE compliant
Expand
View Pricing
Z553778
Scienceware
®
Secador
®
auto-desiccator cabinet,
model 2.0, square profile, clear, AC/DC input 120 V AC
Expand
View Pricing
Z566810
Scienceware
®
Secador
®
auto-desiccator cabinet,
model 4.0, vertical profile, amber, AC/DC input 120 V AC
Expand
View Pricing
Z566799
Scienceware
®
Secador
®
auto-desiccator cabinet,
model 2.0, vertical profile, amber, AC/DC input 120 V AC
Expand
View Pricing
Z553395
Scienceware
®
Secador
®
auto-desiccator cabinet,
model 4.0, horizontal profile, blue end caps, AC/DC input 120 V AC
Expand
View Pricing
Z566802
Scienceware
®
Secador
®
auto-desiccator cabinet,
model 3.0, vertical profile, amber, AC/DC input 120 V AC
Expand
View Pricing