WHA10421057
Whatman® glass microfiber grade GF 92 prefilter discs
white, inorganic binder, 100 ea, 135 mm diam
Synonym(s):
Z750670
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
product name
Whatman® glass microfiber filters with inorganic binder, Grade GF 92, diam. 135 mm
material
borosilicate glass membrane
circles
packaging
pk of 100
manufacturer/tradename
Whatman 10421057
Whatman Article No. 28415412 (US reference)
diam.
135 mm
Looking for similar products? Visit Product Comparison Guide
General description
This filter is used as a membrane prefilter in applications such as the determination of crop protection agent residues by GC or HPLC, in cold sludge determination of beer, in soot separation before gas analyzers and as roll filters in automatic air filtration units.
Features and Benefits
- Inorganic binder provides increased strength
- Temperature resistant up to 500°C
- Medium filtration speed
Other Notes
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Legal Information
Whatman is a registered trademark of Cytiva
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service