WHA10421030
Whatman® glass microfiber grade GF 92 prefilter discs
white, inorganic binder, 200 ea, 50 mm diam
Synonym(s):
Whatman filter, Z698121, fiber filters, glass filter
About This Item
Recommended Products
Product Name
Whatman® glass microfiber filters with inorganic binder, Grade GF 92, diam. 50 mm
material
borosilicate glass membrane
circles
sterility
non-sterile
packaging
pkg of 200 ea
manufacturer/tradename
Whatman 10421030
Whatman Article No. 28415407 (US reference)
diam.
50 mm
Related Categories
General description
Features and Benefits
- Inorganic binder provides increased strength
- Temperature resistant up to 500°C
- Medium filtration speed
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)