Skip to Content
Merck
All Photos(1)

Key Documents

W514500

Sigma-Aldrich

2-Ethylhexyl salicylate

≥99%

Synonym(s):

Octisalate, Octyl salicylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HO)C6H4CO2CH2CH(C2H5)(CH2)3CH3
CAS Number:
Molecular Weight:
250.33
Beilstein:
2730664
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
NACRES:
NA.21
grade:
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

Halal
Kosher

reg. compliance

FDA 21 CFR 117

Assay

≥99%

refractive index

n20/D 1.502 (lit.)

bp

189-190 °C/21 mmHg (lit.)

density

1.014 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

orchid; floral

SMILES string

CCCCC(CC)COC(=O)c1ccccc1O

InChI

1S/C15H22O3/c1-3-5-8-12(4-2)11-18-15(17)13-9-6-7-10-14(13)16/h6-7,9-10,12,16H,3-5,8,11H2,1-2H3

InChI key

FMRHJJZUHUTGKE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

2-Ethylhexyl salicylate is one of the key ingredients (chemical UV filter) of sunscreen lotions due to its ability to absorb ultraviolet radiation.

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Human safety and efficacy of ultraviolet filters and sunscreen products.
Nash JF.
Dermatologic Clinics, 24(1), 35-51 (2006)
UV filters in sunscreen products? a survey.
Rastogi SC.
Contact Dermatitis, 46(6), 348-351 (2002)
M McVean et al.
Molecular carcinogenesis, 24(3), 169-176 (1999-04-16)
Topical application of alpha-tocopherol (alphaTH), the most prominent naturally occurring form of vitamin E, inhibits ultraviolet (UV) B-induced photocarcinogenesis and DNA photodamage in C3H mice in vivo. In this study, we compared alphaTH with other vitamin E compounds and with
P Clarys et al.
Journal of dermatological science, 27(2), 77-81 (2001-09-05)
Temperature influences the stratum corneum adsorption of several topically applied compounds. This study was designed to evaluate the influence of the temperature on the stratum corneum adsorption of 3 UV filters. The UV filters were solubilized in two vehicles, an
Beatriz Albero et al.
Journal of chromatography. A, 1248, 9-17 (2012-06-12)
The determination of organic contaminants in soil is a real challenge due to the large number of these compounds with quite different physico-chemical properties. In the present work, an analytical method was developed for the simultaneous determination in soil of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service