Skip to Content
Merck
All Photos(1)

Key Documents

W355518

Sigma-Aldrich

2-Isopropyl-4-methylthiazole

≥97%, FG

Synonym(s):

Peach thiazole, Tropical thiazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H11NS
CAS Number:
Molecular Weight:
141.23
FEMA Number:
3555
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
15.026
NACRES:
NA.21
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002

Assay

≥97%

refractive index

n20/D 1.5 (lit.)

bp

92 °C/50 mmHg (lit.)

density

1.001 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

green; nutty; fruity; earthy

SMILES string

CC(C)c1nc(C)cs1

InChI

1S/C7H11NS/c1-5(2)7-8-6(3)4-9-7/h4-5H,1-3H3

InChI key

OFLXNHNYPQPQKW-UHFFFAOYSA-N

General description

2-Isopropyl-4-methylthiazole is a sulfur-containing heterocyclic flavor compound that is reported to occur in durian fruit and fresh peach.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

136.4 °F - closed cup

Flash Point(C)

58 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Review of analytical methodologies for volatile nitrogen heterocycles in food.
Pandya RN & Peppard TL.
ACS Symp. Ser., 826, 114?131-114?131 (2002)
Study on Development of a Fresh Peach Flavor
Zhu GY, et al.
Advanced Materials Research, 781-784, 1570-1573 (2013)
Formation and aroma characteristics of heterocyclic compounds in foods.
Ho CT & Carlin JT.
ACS Symp. Ser., 388, 92?104-92?104 (1989)
Sulfur-containing volatiles of durian fruits (Durio zibethinus Murr.).
Weenen H, et al.
Journal of Agricultural and Food Chemistry, 44(10), 3291-3293 (1996)
The role of sulfur compounds in food flavor part I: Thiazoles.
Maga JA & Katz I.
Critical Reviews in Food Science and Nutrition, 6(2), 153-176 (1975)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service