Skip to Content
Merck
All Photos(1)

Key Documents

W342017

Sigma-Aldrich

Damascenone

natural, 1.1-1.4 wt. % (190 proof ethanol), FG

Synonym(s):

β-Damascenone, 1-(2,6,6-Trimethylcyclohexa-1,3-dien-1-yl)-2-buten-1-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H18O
CAS Number:
Molecular Weight:
190.28
FEMA Number:
3420
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
7.108
NACRES:
NA.21

grade

FG
Fragrance grade
Kosher
natural

Quality Level

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 872/2012
FDA 21 CFR 110

form

liquid

concentration

1.1-1.4 wt. % (190 proof ethanol)

color

yellow

refractive index

n20/D 1.350-1.380 (lit.)

density

0.800-0.830 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

apple; smoky; herbaceous; nutty; citrus; woody; rose; wine-like

storage temp.

2-8°C

SMILES string

C\C=C\C(=O)C1=C(C)C=CCC1(C)C

InChI

1S/C13H18O/c1-5-7-11(14)12-10(2)8-6-9-13(12,3)4/h5-8H,9H2,1-4H3/b7-5+

InChI key

POIARNZEYGURDG-FNORWQNLSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Damascenone is a norisoprenoid ketone mainly found in red wines. It occurs naturally in tomato and grapes.

Application

Damascenone is a fragrance ingredient that can be used in shampoos, fine fragrances, decorative cosmetics, toilet soaps as well as non-cosmetic products like household cleaners and detergents.

Biochem/physiol Actions

Taste at 1-5 ppm

Other Notes

Natural occurrence: Apple juice, apricot, black currant, grape, raspberry, strawberry, cognac, rum, whiskey and scotch.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

60.8 °F - closed cup

Flash Point(C)

16 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Frank Benkwitz et al.
Journal of agricultural and food chemistry, 60(25), 6293-6302 (2012-06-06)
In this study three different approaches were employed to identify key odorants in Sauvignon blanc wines. First, the concentrations of the odorants were compared to their respective aroma detection thresholds. The resulting odor activity values (OAV) were transformed into a
J Lalko et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 45 Suppl 1, S192-S198 (2007-11-27)
A toxicologic and dermatologic review of cis-beta-damascone when used as a fragrance ingredient is presented.
Detection and Partial Characterization of Eight. beta.-Damascenone Precursors in Apples (Malus domestica Borkh. Cv. Empire).
Roberts DD, et al.
Journal of Agricultural and Food Chemistry, 42(2), 345-349 (1994)
Lucia Castro-Vázquez et al.
Journal of agricultural and food chemistry, 54(13), 4809-4813 (2006-06-22)
Honeydew honeys from holm-oak, oak, and forest were isolated for aroma compounds by simultaneous distillation-extraction and analyzed by gas chromatography-mass spectrometry. In all, 66 volatile components were identified and quantified. trans-Oak lactone, a characteristic volatile component of oak wood, is
Henryk H Jeleń et al.
Journal of chromatography. A, 1218(42), 7566-7573 (2011-08-13)
Jutrzenka is a sweet liquer wine produced in Poland from the grape variety of the same name, developed in Poland to withstand the harsh climate of winery regions. Jutrzenka wine has a characteristic aroma with strong fruity and flowery notes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service