Skip to Content
Merck
All Photos(1)

Key Documents

W332801

Sigma-Aldrich

2-Acetylthiazole

≥99%, FG

Synonym(s):

1-(1,3-thiazol-2-yl)ethanone, 2- acetyl-1,3-thiazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5NOS
CAS Number:
Molecular Weight:
127.16
FEMA Number:
3328
Beilstein:
109803
EC Number:
Council of Europe no.:
4041
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
15.020
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002

Assay

≥99%

refractive index

n20/D 1.548 (lit.)

bp

89-91 °C/12 mmHg (lit.)

density

1.227 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

hazelnut; nutty; peanut; popcorn; roasted

SMILES string

CC(=O)c1nccs1

InChI

1S/C5H5NOS/c1-4(7)5-6-2-3-8-5/h2-3H,1H3

InChI key

MOMFXATYAINJML-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

2-Acetylthiazole is a volatile flavoring substance generally formed by the Maillard reaction between an amino acid and carbonyl compounds. It is reported to occur in canned sweet corn products, cooked pine mushroom, cooked asparagus and roasted beef.

Application

2-Acetylthiazole can be used as a flavoring agent in food industries. It may be used as a food additive in the preparation of ′fragrant′ rice.

Packaging

Packaged in glass bottles

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

172.4 °F - closed cup

Flash Point(C)

78 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nitrogen-containing heterocyclic compounds identified in the volatile flavor constituents of roasted beef.
Hartman GJ, et al.
Journal of Agricultural and Food Chemistry, 31(5, 1030-1033 (1983)
Formation of flavor components in asparagus. 2. Formation of flavor components in cooked asparagus.
Tressl R, et al.
Journal of Agricultural and Food Chemistry, 25(3), 459-463 (1977)
Jinfeng Peng et al.
Food chemistry, 313, 126136-126136 (2020-01-14)
The adulteration of rice using synthetic aromatic flavorings to fraudulently imitate commercially valuable fragrant rice varieties has attracted extensive attention from regulatory authorities around the world. In order to get convincing evidence of adulteration, appropriate scientific analytical methods need to
Studies on flavor volatiles of some sweet corn products.
Buttery RG, et al.
Journal of Agricultural and Food Chemistry, 42(3), 791-795 (1994)
In Hee Cho et al.
Journal of agricultural and food chemistry, 54(17), 6332-6335 (2006-08-17)
The characteristic aroma-active compounds in raw and cooked pine-mushrooms (Tricholoma matsutake Sing.) were investigated by gas chromatography-olfactometry using aroma extract dilution analysis. 1-Octen-3-one (mushroom-like) was the major aroma-active compound in raw pine-mushrooms; this compound had the highest flavor dilution factor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service