Skip to Content
Merck
All Photos(3)

Key Documents

W316315

Sigma-Aldrich

2-Furyl methyl ketone

greener alternative

natural (US), ≥97%, FG

Synonym(s):

2-Acetylfuran

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6O2
CAS Number:
Molecular Weight:
110.11
FEMA Number:
3163
Beilstein:
107909
EC Number:
Council of Europe no.:
11653
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
13.054
NACRES:
NA.21

grade

FG
Halal
Kosher
natural (US)

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002

Assay

≥97%

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

refractive index

n20/D 1.5070 (lit.)

bp

67 °C/10 mmHg (lit.)

mp

26-28 °C (lit.)

density

1.098 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

greener alternative category

Organoleptic

caramel; cocoa; coffee; brown; nutty; sweet

SMILES string

CC(=O)c1ccco1

InChI

1S/C6H6O2/c1-5(7)6-3-2-4-8-6/h2-4H,1H3

InChI key

IEMMBWWQXVXBEU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Furyl methyl ketone is a volatile heterocyclic flavor compound that may be formed in food due to Maillard reaction. It has been reported in roasted sesame seeds, fried beef and sweet corn products.
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is Biobased and thus aligns with "Less Hazardous Chemical Syntheses" and "Use of Renewable Feedstocks".

Other Notes

Natural occurrence: Burley tobacco, chestnuts, cocoa, coffee, cooked beef and pork, raisins, roasted almonds and peanuts. tamarind, tea, yogurt and wheat bread.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Eye Irrit. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

159.8 °F - closed cup

Flash Point(C)

71 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Shallow-fried beef. Additional flavor components.
Watanabe K & Sato Y.
Journal of Agricultural and Food Chemistry, 20(2), 174-176 (1972)
Amino acid-dependent formation pathways of 2-acetylfuran and 2, 5-dimethyl-4-hydroxy-3 [2H]-furanone in the Maillard reaction
Wang Y, et al.
Food Chemistry, 115(1), 233-237 (2009)
Studies on flavor volatiles of some sweet corn products.
Buttery RG, et al.
Journal of Agricultural and Food Chemistry, 42(3), 791-795 (1994)
Effect of antioxidants on the volatiles of roasted sesame seeds.
Soliman MA, et al.
Journal of Agricultural and Food Chemistry, 33(3), 523-528 (1985)
[Acute hepatitis in subjects exposed to 2-acetylfuran and hydrazine].
G Corsi et al.
La Medicina del lavoro, 74(4), 284-290 (1983-07-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service