Skip to Content
Merck
All Photos(1)

Key Documents

W272809

Sigma-Aldrich

Methyl octanoate

99%, FG

Synonym(s):

Caprylic acid methyl ester, Methyl caprylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)6COOCH3
CAS Number:
Molecular Weight:
158.24
FEMA Number:
2728
Beilstein:
1752270
EC Number:
Council of Europe no.:
398
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.117
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 872/2012
FDA 21 CFR 172.515

Assay

99%

refractive index

n20/D 1.417 (lit.)

bp

194-195 °C (lit.)

density

0.877 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

green; fruity; citrus; sweet

SMILES string

CCCCCCCC(=O)OC

InChI

1S/C9H18O2/c1-3-4-5-6-7-8-9(10)11-2/h3-8H2,1-2H3

InChI key

JGHZJRVDZXSNKQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Methyl octanoate has been identified in the volatile flavor fractions of blue cheese and soursop.

Application


  • Efficient identification of raw and ripe tung oil using headspace GC-MS.: This research demonstrates the use of headspace gas chromatography-mass spectrometry for identifying components, including methyl octanoate, in tung oil. The method offers applications in quality control and authentication of oil products (Xu et al., 2021).

  • Chemical kinetic study of the oxidation of a biodiesel-bioethanol surrogate fuel: methyl octanoate-ethanol mixtures.: The paper investigates the oxidation kinetics of methyl octanoate and ethanol mixtures, providing insights into their use as biofuels. This study is significant for the development of renewable energy sources (Togbe et al., 2010).

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

179.6 °F - closed cup

Flash Point(C)

82 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Volatile flavor components of soursop (Annona muricata).
MacLeod AJ & Pieris NM.
Journal of Agricultural and Food Chemistry, 29(3), 488-490 (1981)
Quantitation, evaluation, and effect of certain microorganisms on flavor components of blue cheese.
Anderson DF & Day EA.
Journal of Agricultural and Food Chemistry, 14(3), 241-245 (1966)
A A Elliott et al.
Biochimica et biophysica acta, 978(2), 337-340 (1989-01-30)
The effects of n-alkanes (n-pentane to n-octane), n-alkanols (n-pentanol to n-undecanol) and two carboxylic esters (methyl pentanoate and methyl octanoate) on the conductance of squid giant axons in a high potassium, zero sodium bathing solution have been examined. Sodium and
C Togbé et al.
The journal of physical chemistry. A, 114(11), 3896-3908 (2010-03-20)
There is a growing interest for using bioethanol-biodiesel fuel blends in diesel engines but no kinetic data and model for their combustion were available. Therefore, the kinetics of oxidation of a biodiesel-bioethanol surrogate fuel (methyl octanoate-ethanol) was studied experimentally in
T J Gianfagna et al.
Life support & biosphere science : international journal of earth space, 5(2), 255-261 (2001-09-07)
Given the constraints on physical space, energy supply, and labor availability, crop production efficiency must be maximized if a sustainable supply of food is to be produced on lunar and planetary space stations. In the modules designed for plant growth

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service