Skip to Content
Merck
All Photos(3)

Key Documents

259942

Sigma-Aldrich

Methyl hexanoate

99%

Synonym(s):

Caproic acid methyl ester, Methyl caproate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4COOCH3
CAS Number:
Molecular Weight:
130.18
Beilstein:
1744683
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.405 (lit.)

bp

151 °C (lit.)

mp

−71 °C (lit.)

solubility

chloroform: soluble 100 mg/mL, clear

density

0.885 g/mL at 25 °C (lit.)

functional group

ester

SMILES string

CCCCCC(=O)OC

InChI

1S/C7H14O2/c1-3-4-5-6-7(8)9-2/h3-6H2,1-2H3

InChI key

NUKZAGXMHTUAFE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hydrodeoxygenation of methyl hexanoate on commercial NiMo/γ-Al2O3 and CoMo/γ-Al2O3 hydro treatment catalysts in either oxide or sulphided form has been investigated[1]. The oxidation of methyl hexanoate in a jet-stirred reactor has been studied[2].

Application

  • Effect of fat nature and aroma compound hydrophobicity on flavor release from complex food emulsions.: This study examines how the type of fat and the hydrophobic nature of aroma compounds, including methyl hexanoate, affect flavor release in food emulsions. The findings highlight the importance of molecular interactions in designing food products with optimized flavor profiles. (Relkin et al., 2004).

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

99.5 °F - closed cup

Flash Point(C)

37.5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hydrodeoxygenation of methyl esters on sulphided NiMo/?-Al2O3 and CoMo/?-Al2O3 catalysts.
Senol OI, et al.
Catalysis Today, 100(3), 331-335 (2005)
Experimental and kinetic modeling study of the oxidation of methyl hexanoate.
Dayma G al.
Energy and Fuels, 22(3), 1469-1479 (2008)
Rebecca M Turner et al.
The Journal of biological chemistry, 289(46), 31837-31845 (2014-10-02)
Insect odorant receptors are heteromeric odorant-gated cation channels comprising a conventional odorant-sensitive tuning receptor (ORx) and a highly conserved co-receptor known as Orco. Orco is found only in insects, and very little is known about its structure and the mechanism
Suhyoung Bahk et al.
BMC biology, 14(1), 83-83 (2016-10-01)
Like most animals, insects rely on their olfactory systems for finding food and mates and in avoiding noxious chemicals and predators. Most insect olfactory neurons express an odorant-specific odorant receptor (OR) along with Orco, the olfactory co-receptor. Orco binds ORs
J C Knight et al.
Journal of the American Mosquito Control Association, 7(1), 37-41 (1991-03-01)
Evidence is reviewed for a dose-dependent reversal of response in mosquito oviposition, in which a compound that attracts or stimulates oviposition may repel or deter oviposition at a higher concentration. On the basis of a review of structure-activity relationships in

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service