W268224
Methyl anthranilate
natural, ≥98%, FG
Synonym(s):
Methyl 2-aminobenzoate, Methyl anthranilate
About This Item
Halal
Kosher
natural
Recommended Products
grade
FG
Halal
Kosher
natural
Agency
meets purity specifications of JECFA
reg. compliance
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
vapor pressure
1 mmHg ( 20 °C)
Assay
≥98%
autoignition temp.
986 °F
refractive index
n20/D 1.582 (lit.)
bp
256 °C (lit.)
mp
24 °C (lit.)
density
1.168 g/mL at 25 °C (lit.)
application(s)
flavors and fragrances
Documentation
see Safety & Documentation for available documents
food allergen
no known allergens
Organoleptic
grape; musty; fruity; sweet; wine-like
SMILES string
COC(=O)c1ccccc1N
InChI
1S/C8H9NO2/c1-11-8(10)6-4-2-3-5-7(6)9/h2-5H,9H2,1H3
InChI key
VAMXMNNIEUEQDV-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
- Functionalization of seawater reverse osmosis membrane with quorum sensing inhibitor to regulate microbial community and mitigate membrane biofouling.: This study investigates the application of methyl anthranilate as a quorum sensing inhibitor in the functionalization of seawater reverse osmosis membranes. The addition of this compound has shown to effectively regulate microbial community dynamics and mitigate membrane biofouling, offering a promising solution to enhance the efficiency and longevity of water treatment systems (Chen et al., 2024).
- Methyl N,N-dimethylanthranilate and ethyl propionate: repellents effective against spotted wing drosophila, Drosophila suzukii.: Research demonstrates that methyl anthranilate, when combined with ethyl propionate, serves as an effective repellent against the pest Drosophila suzukii. This finding is crucial for developing environmentally friendly pest management strategies in agriculture (Conroy et al., 2024).
- Methylanthranilate, a Food Fragrance Attenuates Skin Pigmentation through Downregulation of Melanogenic Enzymes by cAMP Suppression.: This article explores the use of methyl anthranilate in cosmetic formulations, highlighting its efficacy in reducing skin pigmentation by modulating the cellular signaling pathways involved in melanin synthesis. The findings support its potential use in dermatological products aimed at skin lightening and pigmentation correction (Park et al., 2024).
Other Notes
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
262.4 °F - closed cup
Flash Point(C)
128 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service