Skip to Content
Merck
All Photos(1)

Key Documents

W250708

Sigma-Aldrich

Geraniol

≥97%, FCC, FG

Synonym(s):

trans-3,7-Dimethyl-2,6-octadien-1-ol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C=CHCH2CH2C(CH3)=CHCH2OH
CAS Number:
Molecular Weight:
154.25
FEMA Number:
2507
Beilstein:
1722456
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
2.012
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117

vapor density

5.31 (vs air)

vapor pressure

~0.2 mmHg ( 20 °C)

Assay

≥97%

composition

Contains IFRA and EU 1223/2009 restricted Citral (Neral + Geranial) and Geraniol and EU1223/2009 restricted Linalool and Limonene (sum of D, L, and DL)

refractive index

n20/D 1.474 (lit.)

bp

229-230 °C (lit.)

density

0.879 g/mL at 20 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

linalool, citral (neral+geranial), nerol, geraniol, limonene (sum of D, L and DL)

Organoleptic

fruity; citrus; floral; rose; sweet

SMILES string

C\C(C)=C\CC\C(C)=C\CO

InChI

1S/C10H18O/c1-9(2)5-4-6-10(3)7-8-11/h5,7,11H,4,6,8H2,1-3H3/b10-7+

InChI key

GLZPCOQZEFWAFX-JXMROGBWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Geraniol is a monoterpene alcohol that occurs as one of the main flavor constituents in Kansho-shochu, sweet basil, hopped beer and green tea.

Application


  • Alcohol exacerbates psychosocial stress-induced neuropsychiatric symptoms: Attenuation by geraniol.: Investigates the protective effects of geraniol against alcohol-enhanced psychosocial stress, highlighting its potential therapeutic benefits for neuropsychiatric conditions (Ben-Azu et al., 2024).

  • Chemical and Electrophysiological Characterisation of Headspace Volatiles from Yeasts Attractive to Drosophila suzukii.: Studies the attraction of the fruit fly Drosophila suzukii to volatile compounds produced by yeasts, including geraniol, providing insights into pest control strategies (Castellan et al., 2024).


Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

226.4 °F - closed cup

Flash Point(C)

108 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Characteristic flavor of Kamho-shochu (sweet potato spirit).
Ohta T, et al.
Agricultural and Biological Chemistry, 54(6), 1353-1357 (1990)
Biotransformation of hop-derived monoterpene alcohols by lager yeast and their contribution to the flavor of hopped beer.
Takoi K, et al.
Journal of Agricultural and Food Chemistry, 58(8), 5050-5058 (2010)
Characterization of geraniol synthase from the peltate glands of sweet basil.
Iijima Y, et al.
Plant Physiology, 134(1), 370-379 (2004)
Antimicrobial activity of green tea flavor components and their combination effects.
Kubo I, et al.
Journal of Agricultural and Food Chemistry, 40(2), 245-248 (1992)
Gavin L Sacks et al.
Journal of agricultural and food chemistry, 60(12), 2998-3004 (2012-03-09)
1,1,6-Trimethyl-1,2-dihydronaphthalene (TDN) is well-known to contribute "petrol" aromas to aged Riesling wines, but its prevalence and contribution to young Riesling or non-Riesling wines is not well understood. TDN concentrations were measured in 1-3-year-old varietal wines produced from Cabernet franc (n

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service