Skip to Content
Merck
All Photos(4)

Key Documents

W230316

Sigma-Aldrich

Citral

greener alternative

natural, ≥96%, FCC, FG

Synonym(s):

3,7-Dimethyl-2,6-octadienal, Geranial and neral mixture

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C=CHCH2CH2C(CH3)=CHCHO
CAS Number:
Molecular Weight:
152.23
FEMA Number:
2303
Beilstein:
1721871
EC Number:
Council of Europe no.:
109c
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
5.020
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
natural
biological source:
Litsea cubeba Pers.
food allergen:
no known allergens

biological source

Litsea cubeba Pers.

Quality Level

grade

FG
Fragrance grade
Halal
Kosher
natural

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117

vapor density

5 (vs air)

vapor pressure

0.2 mmHg ( 200 °C)

Assay

≥96%

composition

contains IFRA and EU 1223/2009 restricted Citral (Neral + Geranial)

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

refractive index

n20/D 1.488 (lit.)

bp

229 °C (lit.)

density

0.888 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

citral (neral+geranial)

greener alternative category

Organoleptic

fresh; lemon; sweet

SMILES string

[H]C(=O)C=C(C)CC\C=C(\C)C

InChI

1S/C10H16O/c1-9(2)5-4-6-10(3)7-8-11/h5,7-8H,4,6H2,1-3H3

InChI key

WTEVQBCEXWBHNA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives . This product is a Biobased products, showing key improvements in Green Chemistry Principles “Less Hazardous Chemical Syntheses” and “Use of Renewable Feedstock”.

Application


  • Exploring the binding affinity and characteristics of DcitOBP9 in citrus psyllids.: This study delves into the interactions between DcitOBP9, a key odorant-binding protein, and citral in citrus psyllids, providing insights into pest behavior and potential targets for eco-friendly pest management strategies (Pei et al., 2024).

  • Synergistic anti-virulence efficacy of citral and carvacrol against mixed vaginitis causing Candida albicans and Gardnerella vaginalis: An in vitro and in vivo study.: Demonstrates the combined use of citral and carvacrol as a potent anti-virulence strategy against pathogens in mixed vaginitis, showcasing a promising lead for non-antibiotic treatments (Jothi and Gowrishankar, 2024).

  • Effect of polyethoxylated flavonoids (PMFs)-loaded citral and chitosan composite coatings on citrus preservation: From the perspective of fruit resistance.: Discusses innovative food preservation techniques using citral-infused coatings to enhance the storage life and quality of citrus fruits, addressing critical issues in food technology (Li et al., 2024).

  • Synthesis, antifungal activity and action mechanism of novel citral amide derivatives against Rhizoctonia solani.: Explores the synthesis of citral derivatives for use as antifungal agents, offering new solutions for controlling fungal diseases in crops with potential applications in sustainable agriculture (Zhang et al., 2024).

Preparation Note

Extraction method: steam distillation from the fruit of tree.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

208.4 °F - DIN 51758

Flash Point(C)

98 °C - DIN 51758

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lawrence E Marks et al.
Physiology & behavior, 105(2), 443-450 (2011-09-21)
Stimulus context affects judgments of intensity of both gustatory and olfactory flavors, and the contextual effects are modality-specific. Does context also exert separate effects on the gustatory and olfactory components of flavor mixtures? To answer this question, in each of
Xiaoqing Yang et al.
Journal of agricultural and food chemistry, 60(1), 402-409 (2011-12-14)
Multilayer emulsions containing citral were prepared by the layer-by-layer deposition technique based on the electrostatic interaction between negatively charged emulsion droplets and two positively charged biopolymer coatings, chitosan (CS) and ε-polylysine (EPL). The optimum concentrations of both CS and EPL
Jon Lalko et al.
Regulatory toxicology and pharmacology : RTP, 52(1), 62-73 (2008-03-21)
Citral [CAS# 5392-40-5; EINECS# 226-394-6; RIFM # 116; cis- and trans-3,7-dimethyl-2,6-Octadienal] is an important fragrance ingredient appreciated for its powerful lemon-aroma. It is widely used in fragrance formulations and incorporated into numerous consumer products. A comprehensive review of the dermal
Cailin M Rothwell et al.
Neurobiology of learning and memory, 114, 127-140 (2014-06-14)
Retinoic acid, a metabolite of vitamin A, is proposed to play an important role in vertebrate learning and memory, as well as hippocampal-dependent synaptic plasticity. However, it has not yet been determined whether retinoic acid plays a similar role in
Masrat Maswal et al.
Food chemistry, 138(4), 2356-2364 (2013-03-19)
Citral is a flavour component widely used in food and cosmetic industries, but is chemically unstable and degrades over time in aqueous solutions due to acid-catalysed and oxidative reactions leading to loss of desirable flavour. The present study reveals the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service