Skip to Content
Merck
All Photos(1)

Key Documents

T48003

Sigma-Aldrich

Tribromoacetaldehyde

97%

Synonym(s):

Bromal

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Br3CCHO
CAS Number:
Molecular Weight:
280.74
Beilstein:
1098614
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

97%

refractive index

n20/D 1.584 (lit.)

bp

174 °C (lit.)

density

2.665 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

[H]C(=O)C(Br)(Br)Br

InChI

1S/C2HBr3O/c3-2(4,5)1-6/h1H

InChI key

YTGSYRVSBPFKMQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 3 Oral - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

149.0 °F - closed cup

Flash Point(C)

65 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tarek Manasfi et al.
Mutation research, 813, 37-44 (2016-12-25)
Water disinfection treatments result in the formation of disinfection byproducts (DBPs) that have been linked to adverse human health outcomes including higher incidence of bladder and colorectal cancer. However, data about the genotoxicity of DBPs is limited to only a
Sena Kim et al.
Plastic and reconstructive surgery, 138(5), 821e-829e (2016-10-27)
One of the most serious complications of breast surgery using implants is capsular contracture. Several preventive treatments have been introduced; however, the mechanism of capsule formation has not been resolved completely. The authors previously identified negative effects of botulinum toxin
Danae Liviac et al.
Water research, 44(8), 2638-2646 (2010-03-02)
Drinking water contains disinfection byproducts, generated by the interaction of chlorine (or other disinfecting chemicals) with organic matter, anthropogenic contaminants, and bromide/iodide naturally present in most source waters. One class of these chemicals is the halogenated acetaldehydes (HAs), identified in
Olivier Laflamme et al.
Chemosphere, 260, 127660-127660 (2020-08-08)
The occurrence and the fate of 18 ozonation by-products (OBPs) (17 different aldehydes and bromate) were studied over one year in two Canadian drinking water systems. This is the first and only study reporting the occurrence of all these non-halogenated
Gina M Conenello et al.
PLoS pathogens, 3(10), 1414-1421 (2007-10-10)
The proapoptotic PB1-F2 protein of influenza A viruses has been shown to contribute to pathogenesis in the mouse model. Expression of full-length PB1-F2 increases the pathogenesis of the influenza A virus, causing weight loss, slower viral clearance, and increased viral

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service