Skip to Content
Merck
All Photos(1)

Key Documents

T62804

Sigma-Aldrich

1,1,1-Trifluoroacetone

97%

Synonym(s):

α,α,α-Trifluoroacetone, 1,1,1-Trifluoro-2-propanone, 3,3,3-Trifluoroacetone, Methyl trifluoromethyl ketone, Trifluoromethyl methyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCF3
CAS Number:
Molecular Weight:
112.05
Beilstein:
1748614
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

13.62 psi ( 20 °C)

Assay

97%

refractive index

n20/D 1.3 (lit.)

bp

22 °C (lit.)

density

1.252 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(=O)C(F)(F)F

InChI

1S/C3H3F3O/c1-2(7)3(4,5)6/h1H3

InChI key

FHUDAMLDXFJHJE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used in a synthesis of 2-trifluoromethyl-7-azaindoles starting with 2,6-dihalopyridines. The derived chiral imine was used to prepare enantiopure α-trifluoromethyl alanines and diamines via a Strecker reaction followed by either nitrile hydrolysis or reduction.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Flam. Liq. 1 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-22.0 °F - closed cup

Flash Point(C)

-30 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, 251-251 (2007)
Florent Huguenot et al.
The Journal of organic chemistry, 71(18), 7075-7078 (2006-08-26)
Diastereomerically pure alpha-trifluoromethyl alpha-amino nitriles obtained by Strecker-type reactions from chiral CF3 imines and iminium proved to be very attractive versatile intermediates for the synthesis of various alpha-trifluoromethyl amino compounds. From these synthons, both enantiomers of alpha-trifluoromethyl alanine, trifluoromethyl 1,2-diamines
Sheida Esmaielzadeh et al.
Acta chimica Slovenica, 63(2), 351-362 (2016-06-23)
Some cobalt(III) complexes with a potentially tetradentate unsymmetrical NNOS Schiff base ligand have been synthesized and characterized using IR, 1HNMR, UV-Vis spectroscopy and elemental analysis. The equilibrium constants were measured spectrophotometrically for 1:1 adduct formation of the cobalt(III) complexes with
Emre Kinaci et al.
Polymers, 12(9) (2020-09-20)
In this investigation, the terminal double bonds of the side chain epoxidized cardanol glycidyl ether (SCECGE) molecule were further epoxidized in the presence of Oxone® (potassium peroxomonosulfate) and fluorinated acetone. Regular methods for the double bond epoxidation are not effective
Jobst Liebau et al.
The Journal of biological chemistry, 295(29), 9868-9878 (2020-05-22)
Fold-switch pathways remodel the secondary structure topology of proteins in response to the cellular environment. It is a major challenge to understand the dynamics of these folding processes. Here, we conducted an in-depth analysis of the α-helix-to-β-strand and β-strand-to-α-helix transitions

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service