N29600
1,9-Nonanediol
98%
Synonym(s):
Nonamethylene glycol
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
HO(CH2)9OH
CAS Number:
Molecular Weight:
160.25
Beilstein:
1737531
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
98%
form
crystals
bp
177 °C/15 mmHg (lit.)
mp
45-47 °C (lit.)
SMILES string
OCCCCCCCCCO
InChI
1S/C9H20O2/c10-8-6-4-2-1-3-5-7-9-11/h10-11H,1-9H2
InChI key
ALVZNPYWJMLXKV-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 3
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Yuta Toyama et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 197, 148-152 (2018-01-10)
Even-odd alternation of the melting points of α,ω-disubstituted linear alkanes such as alkane-α,ω-diols, alkane-α,ω-dinitriles and α,ω-diaminoalkanes is well known. Melting points for compounds with an even number of carbons in their alkyl chains are systematically higher than those for compounds
Leila Hojabri et al.
Biomacromolecules, 11(4), 911-918 (2010-03-18)
An unsaturated terminal diol, 1,18-octadec-9-endiol (ODEDO), and a saturated terminal diol, 1,9-nonanediol (NDO), were synthesized from oleic acid. The feasibility of utilizing these new diols for the production of thermoplastic polyurethanes (TPUs) was demonstrated by reacting them with a fatty
M Y Ma et al.
Biochemistry, 32(7), 1751-1758 (1993-02-23)
Double-stranded oligodeoxyribonucleotides or single-stranded oligoribonucleotides with specific secondary structure have been proposed as potential antagonists to target nucleic acid-binding proteins (the sense approach). A major limitation of this strategy is that these derivatives are generally considered to be too large
Lucila Navarro et al.
Journal of pharmaceutical sciences, 106(8), 2106-2114 (2017-05-26)
Paclitaxel (PTX) incorporation in poly(lactic-co-glycolic acid) (PLGA) matrices produce films with high tensile rigidity and slow release that fail to deliver the required release rate for most biomedical applications such as in drug eluting stents and cancer treatments. To modify
Kashif Ahmad Ghaffar et al.
Pharmaceutics, 12(2) (2020-02-27)
Triterpenes from the outer bark of birch have many beneficial biological and pharmacological activities. In particular, its wound healing efficacy is of paramount importance. Apart from that, particles of a birch bark dry extract aggregate into a three dimensional network
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service