Skip to Content
Merck
All Photos(1)

Key Documents

405736

Sigma-Aldrich

3-Mercapto-1-propanol

95%

Synonym(s):

3-Mercaptopropanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HS(CH2CH2CH2)OH
CAS Number:
Molecular Weight:
92.16
Beilstein:
1731082
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Assay

95%

refractive index

n20/D 1.495 (lit.)

bp

81-82 °C/10 mmHg (lit.)

density

1.067 g/mL at 20 °C (lit.)

SMILES string

OCCCS

InChI

1S/C3H8OS/c4-2-1-3-5/h4-5H,1-3H2

InChI key

SHLSSLVZXJBVHE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

This compound is used to produce hydrophilic SAMs. Also used as a Traceless Linker for Chemical and Enzymatic Synthesis of Oligosaccharides[1]

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

185.0 °F - closed cup

Flash Point(C)

85 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lisa Crossley et al.
Biosensors, 9(4) (2019-12-11)
We present a low-cost, sensitive and specific DNA field-effect transistor sensor for the rapid detection of a common mutation to the tumour protein 53 gene (TP53). The sensor consists of a commercially available, low-cost, field-effect transistor attached in series to
Mary Beth Wandel et al.
Nature communications, 12(1), 446-446 (2021-01-21)
Complex biological tissues are highly viscoelastic and dynamic. Efforts to repair or replace cartilage, tendon, muscle, and vasculature using materials that facilitate repair and regeneration have been ongoing for decades. However, materials that possess the mechanical, chemical, and resorption characteristics
Fiona C Rowan et al.
ACS chemical biology, 8(10), 2184-2191 (2013-08-09)
Most protein kinases are regulated through activation loop phosphorylation, but the contributions of individual sites are largely unresolved due to insufficient control over sample phosphorylation. Aurora-A is a mitotic Ser/Thr protein kinase that has two regulatory phosphorylation sites on its
Nabyl Merbouh et al.
Organic letters, 9(4), 651-653 (2007-02-09)
The reducing end of protected carbohydrates can be equipped with a series of aglycones via the photochemical installation of a 3-mercaptoethanol linker. This linker is stable during chemical and enzymatic glycosylation reactions but can be activated readily and efficiently to
Chang-Seuk Lee et al.
Talanta, 215, 120899-120899 (2020-04-22)
A simple, label-free and sensitive electrochemical assay is described for the detection of protein kinase A (PKA) activity and inhibition in cancer cell by measuring the change in electrochemical impedance upon phosphorylation. The assay utilized gold nanoparticle (AuNP) and reduced

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service