Skip to Content
Merck
All Photos(1)

Key Documents

N1501

Sigma-Aldrich

Neocuproine

≥98%

Synonym(s):

2,9-Dimethyl-1,10-phenanthroline, DMPHEN

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H12N2
CAS Number:
Molecular Weight:
208.26
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

≥98%

form

crystalline

color

white to beige

SMILES string

Cc1ccc2ccc3ccc(C)nc3c2n1

InChI

1S/C14H12N2/c1-9-3-5-11-7-8-12-6-4-10(2)16-14(12)13(11)15-9/h3-8H,1-2H3

InChI key

IYRGXJIJGHOCFS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Neocuproine is a heterocyclic organic compoundcommonly utilized as a chelating agent in various transition metal-catalyzedreactions. These include alcohol oxidation, oxidative cleavage of carbon-carbonbonds, C-C bond forming coupling reactions, allylation of carbonyls, alkylzincalkynylations, nickel-mediated decarbonylative cross-couplings,Buchwald–Hartwig-type couplings, and allylic substitution reactions.[1]

Application

Neocuproine can be used as a ligand in the:
  • Preparation of fluorinated metal-organic frameworks (F-MOFs).[2]
  • Regioselective C-H arylation of indoles with phenylboronic acids through an oxidative boron Heck reaction.[3]
  • Allylation reaction of aldehydes with allylboronates in the presence of Zn(OH)2 as a catalyst.[4]
  • Iridium-catalyzed silylation of five-membered heteroarenes to synthesize silylarenes.[5]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Assem Barakat et al.
International journal of molecular sciences, 14(12), 23941-23954 (2013-12-20)
[NiCl2(C14H12N2)(H2O)] complex has been synthesized from nickel chloride hexahydrate (NiCl2·6H2O) and 2,9-dimethyl-1,10-phenanthroline (dmphen) as N,N-bidentate ligand. The synthesized complex was characterized by elemental analysis, infrared (IR) spectroscopy, ultraviolet-visible (UV-vis) spectroscopy and differential thermal/thermogravimetric analysis (TG/DTA). The complex was further confirmed
Shū Kobayashi et al.
Chemistry, an Asian journal, 8(9), 2033-2045 (2013-06-19)
Zn(OH)2-catalyzed allylation reactions of aldehydes with allylboronates in aqueous media have been developed. In contrast to conventional allylboration reactions of aldehydes in organic solvents, the α-addition products were obtained exclusively. A catalytic cycle in which the allylzinc species was generated
Mustafa Ozyürek et al.
Analytica chimica acta, 616(2), 196-206 (2008-05-17)
Hydroxyl radicals (OH) generated in the human body may play an important role in tissue injury at sites of inflammation in oxidative stress-originated diseases. As a more convenient, efficient, and less costly alternative to HPLC/electrochemical detection techniques and to the
Saliha Esin Celik et al.
Analytica chimica acta, 674(1), 79-88 (2010-07-20)
A novel on-line HPLC-cupric reducing antioxidant capacity (CUPRAC) method was developed for the selective determination of polyphenols (flavonoids, simple phenolic and hydroxycinnamic acids) in complex plant matrices. The method combines chromatographic separation, constituent analysis, and post-column identification of antioxidants in
Yan Wang et al.
Journal of the American Society for Mass Spectrometry, 19(9), 1353-1360 (2008-07-19)
S-nitrosylation of proteins serves an important role in regulating diverse cellular processes including signal transduction, DNA repair, and neurotransmission. Identification of S-nitrosylation sites is crucial for understanding the significance of this post-translational modification (PTM) in modulating the function of a

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service