Skip to Content
Merck
All Photos(2)

Key Documents

M78801

Sigma-Aldrich

N-Methylpyrrole

99%

Synonym(s):

1-Methyl-1H-pyrrole, N-Methylpyrrole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H7N
CAS Number:
Molecular Weight:
81.12
Beilstein:
104181
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

2.8 (vs air)

vapor pressure

15 mmHg ( 20.2 °C)

Assay

99%

form

liquid

expl. lim.

6 %

bp

112-113 °C (lit.)

mp

−57 °C (lit.)

density

0.914 g/mL at 25 °C (lit.)

SMILES string

Cn1cccc1

InChI

1S/C5H7N/c1-6-4-2-3-5-6/h2-5H,1H3

InChI key

OXHNLMTVIGZXSG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

60.8 °F - closed cup

Flash Point(C)

16 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Giovanni Piani et al.
The journal of physical chemistry. A, 113(52), 14554-14558 (2009-10-16)
We investigated the reaction dynamics of N-methylpyrrole (NMP) along the N-CH3 coordinate, upon excitation energies below 6.4 eV. Ours and previous experiments show clearly the existence of different reaction channels leading to slow and fast fragment production whose relative efficiency
Mingyu Niu et al.
Rapid communications in mass spectrometry : RCM, 20(7), 1077-1081 (2006-02-25)
Different negative fragmentations of synthesized polyamides containing N-methylpyrrole and N-methylimidazole were analyzed by electrospray ionization tandem mass spectrometry with low-energy collision-induced dissociation. An interesting rearrangement derived from amino catalysis of N-methylimidazole was observed. The observation is useful for the study
Gengo Kashiwazaki et al.
Bioorganic & medicinal chemistry, 17(3), 1393-1397 (2009-01-07)
We designed and synthesized alkylating conjugates 5-7 and their partner N-methylpyrrole-N-methylimidazole (PI) polyamides 8, 9. The DNA alkylating activities of conjugates 5-7 were evaluated by high-resolution denaturing polyacrylamide gel electrophoresis with a 219 base pair (bp) DNA fragment containing the
Jiang Zhou et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 11(4), 1157-1162 (2004-12-30)
Electrospray ionization mass spectrometry (ESI-MS) was used to investigate noncovalent complexes formed between four novel polyamides containing N-methylpyrrole (Py) and N-methylimidazole (Im), and human telomeric DNA. Of the four polyamides investigated, PyPyPygammaImImImbetaDp (3) had the highest binding affinity towards the
William Buchmann et al.
Journal of mass spectrometry : JMS, 44(8), 1171-1181 (2009-05-02)
A new method for the determination of the relative affinity of a ligand against various dsDNA sequences is presented by using electrospray ionization time-of-flight mass spectrometry (ESI-QTOF) mass spectrometry. The principle is described here through the complexation of double-stranded DNA

Protocols

Separation of various pyrazines and sulfur compounds including carbon disulfide and dimethyl disulfide.

Separation of various pyrazines and sulfur compounds including carbon disulfide and dimethyl disulfide.

Separation of various pyrazines and sulfur compounds including carbon disulfide and dimethyl disulfide.

Separation of various pyrazines and sulfur compounds including carbon disulfide and dimethyl disulfide.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service