Skip to Content
Merck
All Photos(5)

Key Documents

390836

Sigma-Aldrich

2,4-Dimethylpyrrole

97%

Synonym(s):

2,4-Dimethyl-1-pyrrole, 2,4-Dimethyl-1H-pyrrole, 2,4-Dimethyl-3H-pyrrole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H9N
CAS Number:
Molecular Weight:
95.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.496 (lit.)

bp

165-167 °C (lit.)

density

0.924 g/mL at 25 °C (lit.)

SMILES string

Cc1c[nH]c(C)c1

InChI

1S/C6H9N/c1-5-3-6(2)7-4-5/h3-4,7H,1-2H3

InChI key

MFFMQGGZCLEMCI-UHFFFAOYSA-N

General description

2,4-Dimethylpyrrole is a substituted pyrrole. Its synthesis by using ethyl acetoacetate as starting material has been reported. Its basicity has been evaluated from UV spectral data. Its photodecomposition on irradiation has been reported to afford H2, CH4, C2H6 and polymeric products.

Application

2,4-Dimethylpyrrole may be used in the synthesis of the following:
  • 2,5-bis(2′,4′-dimethyl-5′-pyrryl)p-benzoquinone
  • boron dipyrromethene (BODIPY) dyes
  • 2,4-dimethyl-6-methoxyprodigiosene

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thermal decomposition and ring expansion in 2, 4-dimethylpyrrole. Single pulse shock tube and modeling studies.
Lifshitz A, et al.
The Journal of Physical Chemistry A, 107(24), 4851-4861 (2003)
Photolysis of 2, 5-and 2, 4-dimethylpyrrole vapors at room temperature.
Wu EC and Heicklen J.
Canadian Journal of Chemistry, 50(11), 1678-1689 (1972)
Addition of pyrroles to 1, 4-benzoquinone.
Bullock, E.
Canadian Journal of Chemistry, 36(12), 1744-1744 (1958)
Synthesis and spectral properties of new boron dipyrromethene dyes.
Binchen G, et al.
Dyes and Pigments, 73(2), 206-210 (2007)
The structure of pyrrole salts and the basic strengths of some simple pyrroles.
Bullock, E.
Canadian Journal of Chemistry, 36(12), 1686-1690 (1958)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service