Skip to Content
Merck
All Photos(2)

Documents

I12603

Sigma-Aldrich

β-Ionone

96%

Synonym(s):

beta-Ionone, 4-(2,6,6-Trimethyl-1-cyclohexenyl)-3-buten-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H20O
CAS Number:
Molecular Weight:
192.30
Beilstein:
1909544
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

96%

form

liquid

refractive index

n20/D 1.52 (lit.)

bp

126-128 °C/12 mmHg (lit.)

density

0.945 g/mL at 25 °C (lit.)

SMILES string

CC(=O)\C=C\C1=C(C)CCCC1(C)C

InChI

1S/C13H20O/c1-10-6-5-9-13(3,4)12(10)8-7-11(2)14/h7-8H,5-6,9H2,1-4H3/b8-7+

InChI key

PSQYTAPXSHCGMF-BQYQJAHWSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chang-Hee Kang et al.
Toxicology in vitro : an international journal published in association with BIBRA, 27(2), 782-787 (2012-12-27)
β-Ionone, a precursor of carotenoids, possesses a variety of biological properties such as anti-cancerous, anti-mutagenic and anti-microbial activity. Nevertheless, anti-inflammatory effects of β-ionone remain unknown. In this study, we investigated whether ION attenuates the expression of lipopolysaccharide (LPS)-induced pro-inflammatory mediators
Jonathan T Vogel et al.
Journal of the science of food and agriculture, 90(13), 2233-2240 (2010-07-28)
Tomatoes contain high levels of several carotenoids including lycopene and β-carotene. Beyond their functions as colorants and nutrients, carotenoids are precursors for important volatile flavor compounds. In order to assess the importance of apocarotenoid volatiles in flavor perception and acceptability
Jihai Shao et al.
Aquatic toxicology (Amsterdam, Netherlands), 104(1-2), 48-55 (2011-05-06)
In order to explore the potential targets of toxicity of β-ionone on the photosynthetic system of Microcystis aeruginosa, the polyphasic rise in chlorophyll a (Chl a) fluorescence transient and transcript expression for key genes in photosystem II (PSII) of M.
Vishal Sharma et al.
Bioorganic & medicinal chemistry letters, 22(20), 6343-6346 (2012-09-25)
A series of chalcones (3a-v) have been synthesized by condensation of β-ionone (1) with a variety of aldehydes (2a-v). The synthesized compounds have been screened for their in vitro antimicrobial activity against five bacterial and five fungal strains, using disc
Jianfeng Wang et al.
Investigative ophthalmology & visual science, 53(10), 6355-6369 (2012-08-18)
In human retinal degeneration, rod photoreceptors reactively sprout neurites. The mechanism is unknown in part because of the paucity of animal models displaying this feature of human pathology. We tested the role of cAMP and opsin in sprouting by tiger

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service