Skip to Content
Merck
All Photos(1)

Key Documents

H12900

Sigma-Aldrich

cis-3-Hexen-1-ol

98%

Synonym(s):

(3Z)-3-Hexen-1-ol, (Z)-3-Hexen-1-ol, (Z)-Hex-3-en-1-ol, 3Z-Hexen-1-ol, Blaetteralkohol, cis-1-Hydroxy-3-hexene, cis-3-Hexene-1-ol, cis-3-Hexenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5CH=CHCH2CH2OH
CAS Number:
Molecular Weight:
100.16
Beilstein:
1719712
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

3.45 (vs air)

Assay

98%

form

liquid

color

APHA: ≤100

refractive index

n20/D 1.44 (lit.)

bp

156-157 °C (lit.)

density

0.848 g/mL at 25 °C (lit.)

SMILES string

CC\C=C/CCO

InChI

1S/C6H12O/c1-2-3-4-5-6-7/h3-4,7H,2,5-6H2,1H3/b4-3-

InChI key

UFLHIIWVXFIJGU-ARJAWSKDSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

111.2 °F - closed cup

Flash Point(C)

44 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tsviya Olender et al.
Chemical senses, 37(7), 581-584 (2012-06-15)
Considerable evidence supports the idea that odorant recognition depends on specific sequence variations in olfactory receptor (OR) proteins. Much of this emerges from in vitro screens in heterogenous expression systems. However, the ultimate proof should arise from measurements of odorant
Krista L Ryall et al.
Environmental entomology, 41(3), 648-656 (2012-06-27)
Attraction of emerald ash borer, Agrilus planipennis Fairmaire, to a volatile pheromone was demonstrated in three field experiments using baited green sticky traps. A dose-response curve was generated for male A. planipennis to increasing release rates of (3Z)-dodecen-12-olide ((3Z)-lactone) in
Y Nikaido et al.
Behavioural brain research, 199(2), 241-246 (2008-12-24)
Although various emotional behaviors and activation of the hypothalamic-pituitary-adrenal (HPA) axis of rats are induced by the exposure of 2,5-dihydro-2,4,5-trimethylthiazoline: TMT, a component of fox odor, these odor-induced responses are influenced by the external environment. Our previous study demonstrated that
Sufang Zhang et al.
PloS one, 7(4), e35867-e35867 (2012-05-05)
Exposure of plants to herbivore-induced plant volatiles (HIPVs) alters their resistance to herbivores. However, the whole-genome transcriptional responses of treated plants remain unknown, and the signal pathways that produce HIPVs are also unclear. Time course patterns of the gene expression
Mika Fukada et al.
Chemical senses, 32(6), 633-639 (2007-06-15)
We investigated whether inhalation of green odor (a mixture of equal amounts of trans-2-hexenal and cis-3-hexenol) prevents the skin-barrier disruption induced by chronic restraint stress in rats. To this end, transepidermal water loss (TEWL) was measured as an index of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service