Skip to Content
Merck
All Photos(1)

Key Documents

41163

Supelco

Damascenone

analytical standard

Synonym(s):

β-Damascenone, 1-(2,6,6-Trimethylcyclohexa-1,3-dien-1-yl)-2-buten-1-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H18O
CAS Number:
Molecular Weight:
190.28
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (GC)

form

liquid

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.350-1.380 (lit.)
n20/D 1.510-1.515

density

0.800-0.830 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

C\C=C\C(=O)C1=C(C)C=CCC1(C)C

InChI

1S/C13H18O/c1-5-7-11(14)12-10(2)8-6-9-13(12,3)4/h5-8H,9H2,1-4H3/b7-5+

InChI key

POIARNZEYGURDG-FNORWQNLSA-N

Looking for similar products? Visit Product Comparison Guide

Application

<ul>
<li><strong>Improving Wine Aroma:</strong> Research on enhancing the flavor quality of Laiyang pear wine demonstrated the use of damascenone, which involved sorbitol-utilizing yeasts and co-fermentation strategies to optimize the sensory characteristics of the wine (Deng et al., 2024).</li>
<li><strong>Enhancement of Chinese Baijiu Brewing:</strong> Damascenone′s regulated effects were explored in the context of solid-state fortified Jiuqu and liquid-state fortified agents, enhancing the traditional brewing process of Chinese Baijiu, a significant aspect of cultural heritage and modern beverage technology (Wang et al., 2024).</li>
</ul>

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service