Skip to Content
Merck
All Photos(2)

Key Documents

GF75438445

Vanadium

wire reel, 0.2m, diameter 0.25mm, hard, 99.8%

Synonym(s):

Vanadium, V 005110

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
V
CAS Number:
Molecular Weight:
50.94
MDL number:
UNSPSC Code:
12141749
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

vapor pressure

8 mmHg ( 20 °C)

Assay

99.8%

form

wire

manufacturer/tradename

Goodfellow 754-384-45

resistivity

24.8-26.0 μΩ-cm, 20°C

L × diam.

0.2 m × 0.25 mm

bp

3380 °C (lit.)

mp

1890 °C (lit.)

density

6.11 g/mL at 25 °C (lit.)

SMILES string

[V]

InChI

1S/V

InChI key

LEONUFNNVUYDNQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shinobu Takizawa
Chemical & pharmaceutical bulletin, 57(11), 1179-1188 (2009-11-03)
This review describes our recent efforts in the development of chiral dinuclear vanadium complexes which work as dual activation catalysts for oxidative coupling of 2-naphthols. A chiral dinuclear vanadium(IV) complex (R(a),S,S)-1a possessing (S)-tert-leucine moieties at the 3,3'-positions of the (R)-binaphthyl
Jaclyn M Winter et al.
The Journal of biological chemistry, 284(28), 18577-18581 (2009-04-14)
Nature has developed an exquisite array of methods to introduce halogen atoms into organic compounds. Most of these enzymes are oxidative and require either hydrogen peroxide or molecular oxygen as a cosubstrate to generate a reactive halogen atom for catalysis.
D M Smith et al.
QJM : monthly journal of the Association of Physicians, 101(5), 351-358 (2008-03-06)
To assess the effectiveness of oral vanadium supplementation for glycaemic control in type 2 diabetes by conducting a systematic review of the literature. Eligible studies were identified by searching 14 databases using standardized terms. Experts, study authors and manufacturers were
Mohamad Z Mehdi et al.
Cell biochemistry and biophysics, 44(1), 73-81 (2006-02-04)
Among several metals, vanadium has emerged as an extremely potent agent with insulin-like properties. These insulin-like properties have been demonstrated in isolated cells, tissues, different animal models of type I and type II diabetes as well as a limited number
Lise Coderre et al.
Canadian journal of physiology and pharmacology, 82(10), 833-839 (2004-12-02)
Inorganic and organic compounds of vanadium have been shown to exhibit a large range of insulinomimetic effects in the cardiovascular system, including stimulation of glucose transporter 4 (GLUT-4) translocation and glucose transport in adult cardiomyocytes. Furthermore, administration of vanadium compounds

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service