Skip to Content
Merck
All Photos(2)

Key Documents

GF03408555

Vanadium

wire reel, 0.5m, diameter 0.5mm, as drawn, 99.8%

Synonym(s):

Vanadium, V 005120

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
V
CAS Number:
Molecular Weight:
50.94
MDL number:
UNSPSC Code:
12141749
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

8 mmHg ( 20 °C)

Assay

99.80%

form

wire

manufacturer/tradename

Goodfellow 034-085-55

resistivity

24.8-26.0 μΩ-cm, 20°C

L × diam.

0.5 m × 0.5 mm

bp

3380 °C (lit.)

mp

1890 °C (lit.)

density

6.11 g/mL at 25 °C (lit.)

SMILES string

[V]

InChI

1S/V

InChI key

LEONUFNNVUYDNQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohamad Z Mehdi et al.
Cell biochemistry and biophysics, 44(1), 73-81 (2006-02-04)
Among several metals, vanadium has emerged as an extremely potent agent with insulin-like properties. These insulin-like properties have been demonstrated in isolated cells, tissues, different animal models of type I and type II diabetes as well as a limited number
Birgit Schwenzer et al.
ChemSusChem, 4(10), 1388-1406 (2011-11-22)
Large-scale energy storage has become the main bottleneck for increasing the percentage of renewable energy in our electricity grids. Redox flow batteries are considered to be among the best options for electricity storage in the megawatt range and large demonstration
Dieter Rehder
Journal of inorganic biochemistry, 102(5-6), 1152-1158 (2008-02-08)
In the present focused review, vanadate-dependent haloperoxidases and vanadate-inhibited enzymes which catalyze the hydrolysis of phosphoester bonds are addressed. In these systems, vanadate [HxVO4](3-x)(-) is covalently coordinated to the imidazolyl moiety of an active site histidine, with a geometrical arrangement
D M Smith et al.
QJM : monthly journal of the Association of Physicians, 101(5), 351-358 (2008-03-06)
To assess the effectiveness of oral vanadium supplementation for glycaemic control in type 2 diabetes by conducting a systematic review of the literature. Eligible studies were identified by searching 14 databases using standardized terms. Experts, study authors and manufacturers were
Maja A Larsson et al.
Journal of hazardous materials, 296, 158-165 (2015-04-29)
Blast furnace (BF) slags are commonly applied as soil amendments and in road fill material. In Sweden they are also naturally high in vanadium. The aim of this study was to assess the vanadium bioavailability in BF slags when applied

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service