Skip to Content
Merck
All Photos(2)

Key Documents

GF62540232

Carbon

foil, 25x25mm, thickness 0.25mm, rigid graphite, fine grain size, 99.95%

Synonym(s):

Activated charcoal, C 000410, Charcoal activated

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C
CAS Number:
Molecular Weight:
12.01
MDL number:
UNSPSC Code:
12141908
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

<0.1 mmHg ( 20 °C)

Assay

99.95%

form

foil

autoignition temp.

842 °F

manufacturer/tradename

Goodfellow 625-402-32

resistivity

1375 μΩ-cm, 20°C (graphite)

size × thickness

25 × 25 mm × 0.25 μm

mp

3550 °C (lit.)

SMILES string

[C]

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Formation of Rydberg states in fast ions penetrating thin carbon-foil and gas targets.
Schiwietz et al.
Physical review letters, 59(14), 1561-1564 (1987-10-05)
Systematics of equilibrium charge distributions of ions passing through a carbon foil over the ranges Z=4-92 and E=0.02-6 MeV/u.
Shima et al.
Physical review. A, General physics, 40(7), 3557-3570 (1989-10-01)
Birtcher et al.
Physical review letters, 85(23), 4968-4971 (2000-12-02)
In situ transmission electron microscopy has been used to observe sputtered Au during Xe ion irradiation in transmission geometry. The sputtered Au was collected on an electron transparent carbon foil. Nanoparticles were observed on the collector foil after they were
D J McComas et al.
Proceedings of the National Academy of Sciences of the United States of America, 88(21), 9598-9602 (1991-11-01)
Global imaging of the magnetospheric charged particle population can be achieved by remote measurement of the neutral atoms produced when magnetospheric ions undergo charge exchange with cold exospheric neutral atoms. Previously suggested energetic neutral atom imagers were only able to
Christophe A Marquette et al.
Biosensors & bioelectronics, 19(5), 433-439 (2003-11-19)
A multifunctional bio-sensing chip was designed based on the electrochemiluminescent (ECL) detection of enzymatically produced hydrogen peroxide. Six different oxidases specific for choline, glucose, glutamate, lactate, lysine and urate were non-covalently immobilised on imidodiacetic acid chelating beads (glucose oxidase only)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service