Skip to Content
Merck
All Photos(1)

Key Documents

1.08789

Supelco

Zinc

powder for analysis particle size < 45 µm EMSURE®

Synonym(s):

Zinc

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Zn
CAS Number:
Molecular Weight:
65.39
MDL number:
UNSPSC Code:
12141750
EC Index Number:
231-175-3
NACRES:
NA.21
Assay:
≥95% (complexometric)
form:
powder
Pricing and availability is not currently available.

vapor pressure

1.33 hPa ( 487 °C)

Quality Level

product line

EMSURE®

Assay

≥95% (complexometric)

form

powder

autoignition temp.

460 °C

impurities

≤0.1% Substances insoluble in hydrochloric acid

bp

908 °C/1013 hPa

mp

420 °C

density

7.14 g/cm3 at 20 °C

bulk density

1800‑2700 kg/m3

cation traces

As: ≤0.0001%
Cd: ≤0.05%
Cu: ≤0.005%
Fe: ≤0.005%
Pb: ≤0.005%
Sn: ≤0.001%

storage temp.

2-30°C

SMILES string

[Zn]

InChI

1S/Zn

InChI key

HCHKCACWOHOZIP-UHFFFAOYSA-N

Application

  • ZMYND8 Is a Regulator of Sonic Hedgehog Signaling in ATRA-Mediated Differentiation of Neuroblastoma Cells.: This study reveals the role of ZMYND8, a zinc-binding protein, in regulating Sonic Hedgehog signaling during the differentiation of neuroblastoma cells. This research enhances the understanding of zinc′s involvement in critical cellular processes and its potential in therapeutic applications for neuroblastoma (Guha et al., 2024).
  • Mechanistic insight into the anti-alternaria activity of bimetallic zinc oxide and silver/zinc oxide nanoparticles.: This research provides a detailed mechanistic insight into the antimicrobial properties of bimetallic zinc oxide nanoparticles. The study highlights zinc′s role in developing effective antimicrobial agents, which could be pivotal in addressing fungal infections and improving food safety (Daniel et al., 2024).
  • Bio-fabricated zinc oxide nanoparticles mediated by endophytic fungus Aspergillus sp. SA17 with antimicrobial and anticancer activities: in vitro supported by in silico studies.: The study focuses on the biosynthesis of zinc oxide nanoparticles using endophytic fungi and their dual antimicrobial and anticancer activities. The findings demonstrate the potential of zinc-based nanomaterials in medical applications, particularly in developing new cancer therapies (Abdelrahman et al., 2024).
  • Genome-wide analysis of filamentous temperature-sensitive H protease (ftsH) gene family in soybean.: This article explores the role of zinc in the functioning of the ftsH gene family in soybeans, crucial for plant stress responses and development. The research provides insights into zinc′s role in agricultural biotechnology, enhancing crop resilience and productivity (Wang et al., 2024).

Analysis Note

Assay (complexometric): ≥ 95 %
Identity: passes test
As (Arsenic): ≤ 0.0001 %
Substances insoluble in hydrochloric acid: ≤ 0.1 %
Cd (Cadmium): ≤ 0.05 %
Cu (Copper): ≤ 0.005 %
Fe (Iron): ≤ 0.005 %
Pb (Lead): ≤ 0.005 %
Sn (Tin): ≤ 0.001 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service