Skip to Content
Merck
All Photos(2)

Key Documents

GF30423293

Carbon

rod, 150mm, diameter 2.0mm, graphite, 100%

Synonym(s):

Activated charcoal, C 007911, Charcoal activated

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C
CAS Number:
Molecular Weight:
12.01
MDL number:
UNSPSC Code:
12141908
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

<0.1 mmHg ( 20 °C)

Assay

100%

form

rod

autoignition temp.

842 °F

manufacturer/tradename

Goodfellow 304-232-93

resistivity

1375 μΩ-cm, 20°C (graphite)

size diam.

150 mm × 2.0 mm

mp

3550 °C (lit.)

SMILES string

[C]

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marshall W Bowles et al.
Science (New York, N.Y.), 344(6186), 889-891 (2014-05-09)
Sulfate reduction is a globally important redox process in marine sediments, yet global rates are poorly quantified. We developed an artificial neural network trained with 199 sulfate profiles, constrained with geomorphological and geochemical maps to estimate global sulfate-reduction rate distributions.
Clarissa Gillmann et al.
International journal of radiation oncology, biology, physics, 88(5), 1136-1141 (2014-03-26)
To compare the relative biological effectiveness (RBE)-weighted tolerance doses for temporal lobe reactions after carbon ion radiation therapy using 2 different versions of the local effect model (LEM I vs LEM IV) for the same patient collective under identical conditions.
B Franz Lang et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(16), 5926-5931 (2014-04-09)
Programmed translational bypassing is a process whereby ribosomes "ignore" a substantial interval of mRNA sequence. Although discovered 25 y ago, the only experimentally confirmed example of this puzzling phenomenon is expression of the bacteriophage T4 gene 60. Bypassing requires translational
Elson C Santos et al.
Journal of nanoscience and nanotechnology, 14(6), 4590-4594 (2014-04-18)
The system proposed consists of a flagellum relaxing around a static carbon nanowire to mimics behavior of a natural flagellum moving with damped harmonic motion along a wire under van der Waals and electrostatic forces. This flagellum is composed of
Chi-lai Ho et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 55(5), 749-752 (2014-03-29)
We prospectively compared (11)C-acetate with (18)F-FDG in a PET/CT evaluation of multiple myeloma (MM), specifically on diagnostic accuracy, identification of high-risk patients, and monitoring of treatment response. Dual-tracer PET/CT was performed on 35 pathologically and clinically confirmed and untreated patients

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service