Skip to Content
Merck
All Photos(3)

Documents

E29125

Sigma-Aldrich

2-Ethyl-1,3-hexanediol

97%, Mixture of isomers

Synonym(s):

Ethylhexylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2CH(OH)CH(C2H5)CH2OH
CAS Number:
Molecular Weight:
146.23
Beilstein:
1735324
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

5 (vs air)

Quality Level

Assay

97%

refractive index

n20/D 1.451 (lit.)

bp

241-249 °C (lit.)

mp

−40 °C (lit.)

density

0.933 g/mL at 25 °C (lit.)

SMILES string

CCCC(O)C(CC)CO

InChI

1S/C8H18O2/c1-3-5-8(10)7(4-2)6-9/h7-10H,3-6H2,1-2H3

InChI key

RWLALWYNXFYRGW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

2-Ethyl-1,3-hexanediol (EHD) can be used as:
  • A boron extractant.
  • A reactive solvent in the synthesis of magnetic iron-oxide nanoparticles by non-hydrolytic sol-gel method.
  • A starting material in the selective synthesis of 2-ethyl-1-hydroxy-3-hexanone by oxidation using H2O2.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

276.8 °F - closed cup

Flash Point(C)

136 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Linda Ritzen et al.
Polymers, 13(2) (2021-01-23)
The use of self-healing (SH) polymers to make 3D-printed polymeric parts offers the potential to increase the quality of 3D-printed parts and to increase their durability and damage tolerance due to their (on-demand) dynamic nature. Nevertheless, 3D-printing of such dynamic
The Solvent Extraction of Boron with Synthesized Aliphatic 1, 3?Diols: Stripping and Extraction Behavior of Boron by 2, 2, 5?Trimethyl?1, 3?hexanediol. Solvent extraction and ion exchange
Karakaplan M, et al.
Solvent Extr. Ion Exch., 22(6), 897-911 (2004)
Microwave-assisted oxidation of alcohols using aqueous hydrogen peroxide
Bogda D, et al.
Synlett, 0(1), 143-145 (2000)
2-Ethyl-1,3-hexanediol.
B Ballantyne
Journal of applied toxicology : JAT, 25(3), 248-259 (2005-04-28)
A Izri et al.
Bulletin de la Societe de pathologie exotique (1990), 94(3), 280-283 (2001-10-30)
The authors report the results of a survey on the efficacy against mosquito bites of a repellent, Mousticologne Spécial Zones Infestées (DEET 20%, EHD 15%). Two forms of the product, spray and gel, were tested in Senegal. Repellent efficacy was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service