Skip to Content
Merck
All Photos(2)

Documents

D34108

Sigma-Aldrich

Dibenzylamine

97%

Synonym(s):

(N-Benzylaminomethyl)benzene, Bibenzylamine, DBA, Dibenzylamine (8CI), N,N-Dibenzylamine, N-(Phenylmethyl)benzenemethanamine, N-Benzyl-1-phenylmethanamine, N-Benzylbenzylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5CH2)2NH
CAS Number:
Molecular Weight:
197.28
Beilstein:
909664
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.574 (lit.)

bp

300 °C (lit.)

mp

−26 °C (lit.)

density

1.026 g/mL at 25 °C (lit.)

SMILES string

C(NCc1ccccc1)c2ccccc2

InChI

1S/C14H15N/c1-3-7-13(8-4-1)11-15-12-14-9-5-2-6-10-14/h1-10,15H,11-12H2

InChI key

BWLUMTFWVZZZND-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dibenzylamine is mainly used in the production of rubber accelerators, which play a key role in the vulcanization process. It is used as a precursor in the nitrosative cleavage of tertiary amines, aiding in various chemical reactions. It also serves as an intermediate in the synthesis of palladium complexes.

Application


  • Environmental monitoring in shale gas wastewater: Dibenzylamine was identified among the hazardous substances in shale gas wastewater, with research characterizing its concentration and distribution in the Upper Yangtze River, contributing to improved environmental management practices (Tang et al., 2024).

  • Advancements in organic synthesis: Dibenzylamine was used in a novel synthetic strategy for (L)-Monomethyl Tyrosine via bulky ′forced-traceless′ regioselective Pd-catalyzed C(sp(2))-H activation, showcasing its utility in pharmaceutical compound development (Illuminati et al., 2023).

  • Application in crystallography: The crystal structure of di-benzyl-ammonium was elucidated, providing insights into molecular interactions and potential applications in material science and drug design (Traoré et al., 2023).

  • Utilization in green chemistry: Dibenzylamine facilitated a green approach towards Triazole forming reactions, aiming to develop anticancer drugs by minimizing environmental impact and enhancing reaction efficiency (Rastogi et al., 2023).


Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1C

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

289.4 °F - closed cup

Flash Point(C)

143 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thermal decomposition of the rubber vulcanization agent, zinc dibenzyldithiocarbamate, and its potential role in nitrosamine formation in hams processed in elastic nettings
Helmick et al.
Journal of Agricultural and Food Chemistry, 42, 2541-2544 (1994)
Palladium-Assisted Formation of Carbon- Carbon Bonds. 8.1 Synthesis and Reactivity toward Internal Alkynes, Carbon Monoxide, and Isocyanides of Orthopalladated Dibenzylamine Complexes
Vicente et al.
Organometallics, 18, 2683-2693 (1999)
Nitrosative cleavage of tertiary amines
Smith et al.
Journal of the American Chemical Society, 89, 1147-1157 (1967)
Sean D Donevan et al.
Epilepsia, 44(10), 1274-1279 (2003-09-27)
Previously we demonstrated that L-(+)-beta-hydroxybutyrate (L-BHB), acetoacetate (ACA), acetone, and dibenzylamine (DBA) were anticonvulsant in an audiogenic seizure-susceptible model, and that DBA was a bioactive contaminant identified in commercial lots of L-BHB. In the present study, we asked whether these
D W Thompson
Journal of pharmaceutical sciences, 71(5), 536-538 (1982-05-01)
A GLC method is described for the determination of iminodibenzyl and desipramine impurities in imipramine hydrochloride and its formulated products. These impurities were extracted from an alkaline solution with a mixture of 30% methylene chloride in hexane for chromatography on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service