Skip to Content
Merck
All Photos(3)

Key Documents

471321

Sigma-Aldrich

2-(Diethylamino)ethanol

≥99.5%

Synonym(s):

N,N-Diethylethanolamine, DEAE, DEEA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)2NCH2CH2OH
CAS Number:
Molecular Weight:
117.19
Beilstein:
741863
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

4.04 (vs air)

vapor pressure

1 mmHg ( 20 °C)

Assay

≥99.5%

expl. lim.

11.7 %

refractive index

n20/D 1.441 (lit.)

bp

161 °C (lit.)

density

0.884 g/mL at 25 °C (lit.)

SMILES string

CCN(CC)CCO

InChI

1S/C6H15NO/c1-3-7(4-2)5-6-8/h8H,3-6H2,1-2H3

InChI key

BFSVOASYOCHEOV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-(Diethylamino)ethanol (DEEA) is a tertiary alkanolamine multi-component aqueous solvent. It has a high chemical stability and resistance against degradation. DEEA is used to prepare quaternary ammonium salts. These salts are widely used as phase transfer catalysts to promote reactions between immiscible phases.

Application

2-(Diethylamino)ethanol (DEEA) can be used as a co-solvent with methyldiethanolamine (MDEA) and sulfolane to investigate the CO2 absorption and desorption behavior in aqueous solutions. Additionally, DEAE is used to prepare N-substituted glycine derivatives and these compounds are used in the synthesis of peptides and proteins.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

122.0 °F - closed cup

Flash Point(C)

50 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Glucamines as green alternatives to conventional amino alcohols in metalworking fluids
Seddig, et al.
Materials and Corrosion, 74, 394-402 (2023)
Amino alcohol additives for the fast living radical polymerization of methyl methacrylate with RuCl2 (PPh3)3.
Ando T, et al.
Journal of Polymer Science Part A: Polymer Chemistry, 41(22), 3597-3605 (2003)
CO2 absorption/desorption in aqueous DEEA/MDEA and their hybrid solutions with sulfolane
Nozaeim, et al.
Journal of Natural Gas Science and Engineering, 76, 103219-103219 (2020)
Kinetics of carbon dioxide reaction with diethylaminoethanol in aqueous solutions.
Kim CJ and Savage DW.
Chemical Engineering Science, 42(6), 1481-1487 (1987)
Probing molecular interactions in the system $\{$water+ 2-(diethylamino) ethanol$\}$ using solvatochromic dyes and excess partial molar enthalpies, at 298.15 K
Moita, et al.
Journal of Molecular Liquids, 366, 120264-120264 (2022)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service