Skip to Content
Merck
All Photos(2)

Key Documents

D65658

Sigma-Aldrich

Dichloromethyl methyl ether

98%

Synonym(s):

α,α−Dichloromethyl methyl ether, 1,1-Dichlorodimethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2CHOCH3
CAS Number:
Molecular Weight:
114.96
Beilstein:
1900293
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.431 (lit.)

bp

85 °C (lit.)

density

1.271 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

COC(Cl)Cl

InChI

1S/C2H4Cl2O/c1-5-2(3)4/h2H,1H3

InChI key

GRTGGSXWHGKRSB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Common formylating reagent for aromatic compounds, chlorinating agent for carboxylic acids, and reactant with borinic esters for the preparation of alkenones.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

107.6 °F - closed cup

Flash Point(C)

42 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Asymmetry, 4, 2183-2183 (1993)
The Journal of Organic Chemistry, 57, 6847-6847 (1992)
Synlett, 626-626 (1992)
Dong Yu Zhu et al.
Nanoscale, 10(39), 18651-18656 (2018-09-29)
Multidrug-resistant biofilms are highly resistant to current antimicrobial therapies. We have developed an antimicrobial platform that integrates the bacteria-killing phytochemical carvacrol into dynamically crosslinked polymer nanocomposites (DCPNs). Taking advantage of a reversibly crosslinked Schiff-base scaffold throughout the engineered emulsions, DCPNs

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service