Skip to Content
Merck
All Photos(2)

Documents

D47102

Sigma-Aldrich

Dibutyl maleate

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3OCOCH=CHCOO(CH2)3CH3
CAS Number:
Molecular Weight:
228.28
Beilstein:
1726634
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

96%

form

liquid

refractive index

n20/D 1.445 (lit.)

bp

281 °C (lit.)

density

0.988 g/mL at 25 °C (lit.)

SMILES string

[H]\C(=C(/[H])C(=O)OCCCC)C(=O)OCCCC

InChI

1S/C12H20O4/c1-3-5-9-15-11(13)7-8-12(14)16-10-6-4-2/h7-8H,3-6,9-10H2,1-2H3/b8-7-

InChI key

JBSLOWBPDRZSMB-FPLPWBNLSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Sens. 1 - STOT RE 2 Oral

Target Organs

Kidney

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

248.0 °F

Flash Point(C)

120 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Allergic contact dermatitis from occupational exposure to dibutyl maleate.
Denis Sasseville et al.
Contact dermatitis, 60(5), 298-300 (2009-04-29)
J Thormann et al.
Contact dermatitis, 13(5), 314-316 (1985-11-01)
10 of 20 workers in an envelope-making factory developed contact dermatitis. The cause was dibutyl maleinate, which was added to a polyvinylacetate glue. Dibutyl maleinate is a strong sensitizer and should be withdrawn from industrial use.
Alfredo A Marchetti et al.
Chemosphere, 52(5), 861-868 (2003-05-22)
The addition of oxygen-bearing compounds to diesel fuel considerably reduces particulate emissions. TGME and DBM have been identified as possible diesel additives based on their physicochemical characteristics and performance in engine tests. Although these compounds will reduce particulate emissions, their
Xiaoping Zhan et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 66(3), 429-434 (2007-01-02)
Poly(2-hydroxy-3-phenoxypropylacrylate, 4-hydroxybutyl acrylate, dibutyl maleate) membrane was synthesized by UV curing method in our laboratory for the first time. When above-mentioned monomers were in the weight ratio of 4:4:2, the membrane not only had perfect permeation property but also had
Contact dermatitis from dibutyl maleate.
J S English et al.
Contact dermatitis, 13(5), 337-338 (1985-11-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service