Skip to Content
Merck
All Photos(2)

Key Documents

D1928

Sigma-Aldrich

Fenaminosulf

≥98%

Synonym(s):

4-Dimethylaminobenzenediazosulfonic acid sodium salt, p-Diazodimethylaniline sodium sulfonate, Sodium [4-(dimethylamino)phenyl]diazenesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C8H10N3O3SNa
CAS Number:
Molecular Weight:
251.24
Beilstein:
4170376
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98%

form

powder

storage temp.

−20°C

SMILES string

[Na+].CN(C)c1ccc(cc1)N=NS([O-])(=O)=O

InChI

1S/C8H11N3O3S.Na/c1-11(2)8-5-3-7(4-6-8)9-10-15(12,13)14;/h3-6H,1-2H3,(H,12,13,14);/q;+1/p-1

InChI key

IWDQPCIQCXRBQP-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Used in biological and pharmacological studies including:
  • Cloning and characterization of GABA transporter
  • Activity of anti-RSV carbonucleosides
  • Inhibition by fungicides of urea hydrolysis and nitrification of urea nitrogen in soil

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Chronic 3

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tao Hu et al.
Anticancer research, 35(9), 4655-4663 (2015-08-09)
Esophageal squamous cell carcinoma (ESCC) is a recalcitrant cancer. Mouse models of this disease could be used for discovery of more effective therapy for ESCC. The green fluorescent protein (GFP)-expressing human esophageal cancer EC1 cell line was established with a
P Stojanov et al.
Pacing and clinical electrophysiology : PACE, 21(1 Pt 1), 65-68 (1998-02-25)
Endocardial pacing system implantation has been performed in 15 children of mean age 37 months (ranging from 1 day to 89 months). Endocardial lead fixation was performed by means of slowly resorbable suture (Dexon) to allow spontaneous lead migration as
S Vainionpää et al.
Biomaterials, 8(1), 46-48 (1987-01-01)
The initial shear strength and changes in flexural strength of self-reinforced, absorbable polyglycolide (PGA) composite rods, submerged in distilled water (at 37 degrees C) for a period of 6 wk, were investigated. The recently developed self-reinforced absorbable material consists of
M Shun et al.
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials, 20(2), 56-58 (1997-02-01)
This paper reports the effect on nymexazol protecting the Rhizoctonia solani of Panax ginseng. The protective effect is in 80-95%. The drug can substitute quintozene and fenaminosulf in production.
S B Pai
Mutation research, 119(3), 289-291 (1983-03-01)
Fenaminosulf (p-dimethylaminobenzenediazo sodium sulfonate, CAS registry No. 140-56-7) which is an active ingredient in several commercial fungicides was reported to be mutagenic in Salmonella typhimurium (McCann et al., 1975), Bacillus subtilis (Kada et al., 1974) and shown to cause chromosome

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service