Skip to Content
Merck
All Photos(3)

Documents

D161802

Sigma-Aldrich

N,N′-Dimethylhydrazine dihydrochloride

98%

Synonym(s):

1,2-Dimethylhydrazine dihydrochloride, N,N′-Dimethylhydrazine (sym.) dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3NHNHCH3 · 2HCl
CAS Number:
Molecular Weight:
133.02
Beilstein:
3905684
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

crystals

mp

167-169 °C (dec.) (lit.)

SMILES string

Cl[H].Cl[H].CNNC

InChI

1S/C2H8N2.2ClH/c1-3-4-2;;/h3-4H,1-2H3;2*1H

InChI key

WLDRFJDGKMTPPV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Induces colon cancer in rats and mice.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hisako Hori et al.
Regulatory toxicology and pharmacology : RTP, 112, 104578-104578 (2020-01-15)
Genotoxicity occurring at the target organs of carcinogenesis is important for understanding the mechanisms of chemical carcinogenicity and also for setting of threshold estimation. In vivo gene mutations have been evaluated by transgenic animal models in which any organ can
Luciana Azevedo et al.
Journal of the science of food and agriculture, 99(2), 624-631 (2018-06-29)
Camu-camu (Myrciaria dubia) is a typical Amazonian fruit and has high antioxidant capacity due to its high levels of vitamin C and phenolic compounds. This study aimed to determine the phytochemicals, antioxidant capacity and antimutagenic effects of camu-camu fruits with
Leila Kaeid Sharaf et al.
Nutrition and cancer, 70(6), 946-955 (2018-09-06)
Epidemiological and experimental observations have shown that nonsteroidal anti-inflammatory drugs especially selective cyclooxygenase-2 (COX-2) inhibitors and probiotics reduce the incidence risk of colon cancer. Therefore, the present study was designed to assess the prophylactic potentials of probiotics (Lactobacillus acidophilus and
Preety Ghanghas et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 78, 239-247 (2016-02-24)
Non-steroidal anti-inflammatory drugs (NSAIDs) are emerging as novel chemopreventive agents against a variety of cancers owing to their capability in blocking the tumor development by cellular proliferation and by promoting apoptosis. Inflammation is principal cause of colon carcinogenesis. A missing
Vijay L Kumar et al.
Drug development research, 80(8), 1089-1097 (2019-09-01)
Anti-inflammatory drugs are well known to reduce the risk of colon cancer and prophylactic use of such agents is gaining acceptance as a cancer prevention therapy. As artesunate, an antimalarial drug, has been shown to exhibit chemopreventive properties, the present

Articles

Cancer research has revealed that the classical model of carcinogenesis, a three step process consisting of initiation, promotion, and progression, is not complete.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service