Skip to Content
Merck
All Photos(3)

Key Documents

D161403

Sigma-Aldrich

5,5-Dimethylhydantoin

97%

Synonym(s):

Fennosurf 300, 4,4-Dimethyl-2,5-dioxoimidazolidine, DMH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H8N2O2
CAS Number:
Molecular Weight:
128.13
Beilstein:
2827
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

crystals

mp

174-177 °C (lit.)

SMILES string

CC1(C)NC(=O)NC1=O

InChI

1S/C5H8N2O2/c1-5(2)3(8)6-4(9)7-5/h1-2H3,(H2,6,7,8,9)

InChI key

YIROYDNZEPTFOL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for synthesis of:
  • N-chloroheterocyclic antimicrobials
  • β-Amino alcohols as inhibitors of antitubercular target N-acetyltransferase
  • Second-generation selective inhibitors of hepatitis C virus NS3 serine protease
  • Rechargeable biocidal cellulose
  • N-chlorohydantoins

Reactant for asymmetric chlorolactonization reactions

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Umit Makal et al.
Biomaterials, 27(8), 1316-1326 (2005-09-27)
Polyurethanes (PUs) with soft blocks containing semifluorinated (-CH2OCH2CF3) and 5,5-dimethylhydantoin pendant groups were prepared and employed (2 wt%) as biocidal polymeric surface modifiers (PSMs) for a conventional PU coating comprised of an isophorone diisocyanate/1,4-butanediol-derived hard block (50%) and poly(tetramethylene oxide)
A C de Groot et al.
Contact dermatitis, 18(4), 197-201 (1988-04-01)
The relationship between contact allergy to formaldehyde and positive patch test reactions to DMDM hydantoin was investigated. 35 formaldehyde-allergic patients were patch tested with serial dilutions of formaldehyde (0.1%-0.3%-1.0% aq.) and DM hydantoin (the non-formaldehyde-containing parent compound of DMDM hydantoin).
Ana S Moreira et al.
Virulence, 8(6), 782-796 (2016-09-23)
During long-term lung infection in cystic fibrosis (CF) patients, Burkholderia cenocepacia faces multiple selective pressures in this highly stressful and fluctuating environment. As a consequence, the initial infecting strain undergoes genetic changes that result in the diversification of genotypes and
Nicholas J Hunt et al.
Neurobiology of aging, 36(1), 292-300 (2014-09-13)
Animal studies have shown that decreased orexin expression changes sleep regulation with normal aging. This study examined orexin A and B expression in the tuberal hypothalamus in infants (0-1 year; n = 8), children (4-10 years; n = 7), young
Joseph Jacob et al.
BMC medicine, 14(1), 190-190 (2016-11-24)
To evaluate computer-based computer tomography (CT) analysis (CALIPER) against visual CT scoring and pulmonary function tests (PFTs) when predicting mortality in patients with connective tissue disease-related interstitial lung disease (CTD-ILD). To identify outcome differences between distinct CTD-ILD groups derived following

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service