D138606
3,3-Dimethylacrylic acid
97%
Synonym(s):
3-Methyl-2-butenoic acid, 3,3-Dimethylacrylic acid, 3-Methylcrotonic acid, Senecioic acid
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Linear Formula:
(CH3)2C=CHCOOH
CAS Number:
Molecular Weight:
100.12
Beilstein:
1720305
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
97%
form
solid
bp
194-195 °C (lit.)
mp
65-70 °C (lit.)
SMILES string
C\C(C)=C\C(O)=O
InChI
1S/C5H8O2/c1-4(2)3-5(6)7/h3H,1-2H3,(H,6,7)
InChI key
YYPNJNDODFVZLE-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
200.1 °F - closed cup
Flash Point(C)
93.4 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
K Roth et al.
The Journal of pediatrics, 88(2), 229-235 (1976-02-01)
A patient is described in whom lactic acidosis of very severe degree was found to coincide with the presence of beta-methylcrotonic acid and rho-hydroxyphenyllactic acid in urine in large amounts, while beta-hydroxyisovaleric acid was found to be a relatively minor
Hirofumi Inoue et al.
Biological & pharmaceutical bulletin, 42(5), 845-849 (2019-05-08)
It has been suggested that imbalances in gut microbiota are related to diseases associated with metabolism, the central nervous system, etc. Therefore, analysis of short-chain fatty acids (SCFAs) produced by gut microbiota is very important as an indicator of causation
Nareshbabu Kamatham et al.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology, 18(10), 2411-2420 (2019-07-28)
ortho-Nitrobenzyl (oNB) triggers have been extensively used to release various molecules of interest. However, the toxicity and reactivity of the spent chromophore, o-nitrosobenzaldehyde, remains an unaddressed difficulty. In this study we have applied the well-established supramolecular photochemical concepts to retain
M Duran et al.
Clinica chimica acta; international journal of clinical chemistry, 90(2), 187-193 (1978-12-01)
The organic acid excretion in a patient with 3-hydroxy-3-methylglutaryl-CoA lyase deficiency is described. In addition to 3-hydroxy-3-methylglutaric, 3-methylglutaconic, 3-methylglutaric and 3-hydroxyisovaleric acids, substantial amounts of glutaric and adipic acids were repeatedly observed. 3-Methylcrotonic acid and possibly one of its isomers
Shogo Inamine et al.
Bioorganic & medicinal chemistry, 22(21), 5902-5909 (2014-10-07)
IsoVa-RYYRIK-NH2 is a highly specific antagonist ligand of the opioid receptor-like 1 (ORL1) receptor, an endogenous ligand of which is 17-mer peptide nociceptin. ORL1 antagonists have potential for clinical use as analgesic and antineuropathic drugs, and thus information on the
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service