All Photos(1)
About This Item
Linear Formula:
(HOC6H4)2CO
CAS Number:
Molecular Weight:
214.22
Beilstein:
1874572
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
Assay
99%
form
powder
mp
213-215 °C (lit.)
SMILES string
Oc1ccc(cc1)C(=O)c2ccc(O)cc2
InChI
1S/C13H10O3/c14-11-5-1-9(2-6-11)13(16)10-3-7-12(15)8-4-10/h1-8,14-15H
InChI key
RXNYJUSEXLAVNQ-UHFFFAOYSA-N
Gene Information
rat ... Ar(24208)
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Irene Gómez Pinto et al.
Journal of medicinal chemistry, 51(22), 7205-7215 (2008-10-28)
The human ribonucleoprotein telomerase is a validated anticancer drug target, and hTR-P2b is a part of the human telomerase RNA (hTR) essential for its activity. Interesting ligands that bind hTR-P2b were identified by iteratively using a tandem structure-based approach: docking
Soizic Prado et al.
Bioorganic & medicinal chemistry, 15(5), 2177-2186 (2007-01-09)
We recently reported that 3,3-dimethyl-3H-benzofuro[3,2,f][1]-benzopyran and its hydrogenated analogue are selective in vitro inhibitors of mycobacterial growth. However, their lack of in vivo activity on a murine model of Mycobacterium tuberculosis infection due to their poor bioavailability led to a
Daniel Molins-Delgado et al.
Environmental science & technology, 51(19), 10983-10990 (2017-09-06)
The present study uses bird eggs of seven wild species as a biomonitoring tool for sunscreens occurrence. Seven UV filters (UV-Fs), including 3 hydroxy-metabolites of oxybenzone (benzophenone 3, BP3) were characterized in unhatched eggs from Doñana Natural Space (Spain). High
Larissa M Podust et al.
Antimicrobial agents and chemotherapy, 51(11), 3915-3923 (2007-09-12)
Sterol 14alpha-demethylase (CYP51), a major checkpoint in membrane sterol biosynthesis, is a key target for fungal antibiotic therapy. We sought small organic molecules for lead candidate CYP51 inhibitors. The changes in CYP51 spectral properties following ligand binding make CYP51 a
Markus A Lill et al.
Journal of medicinal chemistry, 48(18), 5666-5674 (2005-09-02)
We investigated the influence of induced fit of the androgen receptor binding pocket on free energies of ligand binding. On the basis of a novel alignment procedure using flexible docking, molecular dynamics simulations, and linear-interaction energy analysis, we simulated the
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service