Merck
  • Impact of induced fit on ligand binding to the androgen receptor: a multidimensional QSAR study to predict endocrine-disrupting effects of environmental chemicals.
Impact of induced fit on ligand binding to the androgen receptor: a multidimensional QSAR study to predict endocrine-disrupting effects of environmental chemicals.
Journal of medicinal chemistry (2005-09-02)
Markus A Lill, Fabienne Winiger, Angelo Vedani, Beat Ernst
ABSTRACT
We investigated the influence of induced fit of the androgen receptor binding pocket on free energies of ligand binding. On the basis of a novel alignment procedure using flexible docking, molecular dynamics simulations, and linear-interaction energy analysis, we simulated the binding of 119 molecules representing six compound classes. The superposition of the ligand molecules emerging from the combined protocol served as input for Raptor, a receptor-modeling tool based on multidimensional QSAR allowing for ligand-dependent induced fit. Throughout our study, protein flexibility was explicitly accounted for. The model converged at a cross-validated r(2) = 0.858 (88 training compounds) and yielded a predictive r(2) = 0.792 (26 test compounds), thereby predicting the binding affinity of all compounds close to their experimental value. We then challenged the model by testing five molecules not belonging to compound classes used to train the model: the IC(50) values were predicted within a factor of 4.5 compared to the experimental data. The demonstrated predictivity of the model suggests that our approach may well be beneficial for both drug discovery and the screening of environmental chemicals for endocrine-disrupting effects, a problem that has recently become a cause for concern among scientists, environmental advocates, and politicians alike.
Product Description
SAFC
Cholesterol, Plant-Derived, SyntheChol®
Supelco
Benzophenone, analytical standard
Supelco
4,4′-DDT solution, certified reference material, 5000 μg/mL in methanol
Sigma-Aldrich
Bisphenol A, 97%
Sigma-Aldrich
Benzophenone, purified by sublimation, ≥99%
Supelco
Bisphenol A, ≥99%
Sigma-Aldrich
3-Chlorophenol, 98%
Sigma-Aldrich
4,4′-Dihydroxybenzophenone, 99%
Sigma-Aldrich
4-(Heptyloxy)phenol, 97%
Sigma-Aldrich
4-Hydroxybenzophenone, 98%
Sigma-Aldrich
Nonylphenol, technical grade, mixture of ring and chain isomers
Sigma-Aldrich
Methyl 4-hydroxybenzoate, tested according to Ph. Eur.
Sigma-Aldrich
Propyl 4-hydroxybenzoate, ≥99%
Sigma-Aldrich
4-Cumylphenol, 99%
Sigma-Aldrich
trans-Chalcone, 97%
Supelco
Dexamethasone, VETRANAL®, analytical standard
Supelco
Nonylphenol, PESTANAL®, analytical standard, technical mixture
Supelco
4,4′-DDD, PESTANAL®, analytical standard
Sigma-Aldrich
Benzophenone, ReagentPlus®, 99%
Sigma-Aldrich
Methyl 4-hydroxybenzoate, ≥99%, FCC
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.
Sigma-Aldrich
Propyl 4-hydroxybenzoate, tested according to Ph. Eur.
Sigma-Aldrich
Methyl 4-hydroxybenzoate, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Methyl 4-hydroxybenzoate, ReagentPlus®, ≥99.0%, crystalline
Sigma-Aldrich
Methyl 4-hydroxybenzoate, analytical standard
Sigma-Aldrich
Propyl 4-hydroxybenzoate, BioXtra
Sigma-Aldrich
Estradiol, meets USP testing specifications
Supelco
Testosterone solution, 1.0 mg/mL in 1,2-dimethoxyethane, drug standard
Sigma-Aldrich
Hydrocortisone solution, 50 μM, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Methyl 4-hydroxybenzoate, BioXtra, ≥99.0% (titration)