Skip to Content
Merck
All Photos(1)

Key Documents

C8527

Sigma-Aldrich

Chlorhexidine dihydrochloride

≥98%

Synonym(s):

1,6-Bis(N5-[p-chlorophenyl]-N1-biguanido)hexane; 1,1′-Hexamethylenebis(5-[p-chlorophenyl]biguanide), Lisium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H30Cl2N10 · 2HCl
CAS Number:
Molecular Weight:
578.37
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98%

form

powder

storage temp.

2-8°C

SMILES string

Cl.Clc1ccc(NC(=N)NC(=N)NCCCCCCNC(=N)NC(=N)Nc2ccc(Cl)cc2)cc1

InChI

1S/C22H30Cl2N10.2ClH/c23-15-5-9-17(10-6-15)31-21(27)33-19(25)29-13-3-1-2-4-14-30-20(26)34-22(28)32-18-11-7-16(24)8-12-18;;/h5-12H,1-4,13-14H2,(H5,25,27,29,31,33)(H5,26,28,30,32,34);2*1H

InChI key

WJLVQTJZDCGNJN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C M Martín-Navarro et al.
Graefe's archive for clinical and experimental ophthalmology = Albrecht von Graefes Archiv fur klinische und experimentelle Ophthalmologie, 251(9), 2111-2117 (2013-05-21)
Acanthamoeba is an opportunistic pathogen which is the causal agent of a sight-threatening ulceration of the cornea known as "Acanthamoeba keratitis" (AK) and, more rarely, an infection of the central nervous system called "granulomatous amoebic encephalitis" (GAE). The symptoms of
Leighann Sherry et al.
BMC oral health, 13, 47-47 (2013-09-26)
A number of oral diseases, including periodontitis, derive from microbial biofilms and are associated with increased antimicrobial resistance. Despite the widespread use of mouthwashes being used as adjunctive measures to control these biofilms, their prolonged use is not recommended due
Afshin Haraji et al.
Journal of orofacial pain, 27(3), 256-262 (2013-07-25)
To assess the effects of intra-alveolar application of chlorhexidine gel on the incidence of alveolar osteitis (dry socket) and the severity of postsurgical pain. A total of 160 impacted mandibular third molars were extracted in 80 patients enrolled in this
L Evans et al.
Anaesthesia, 68(12), 1243-1246 (2013-10-12)
Chlorhexidine contamination of equipment used in central neuraxial anaesthesia has been implicated in causing adhesive arachnoiditis. We measured the extent of chlorhexidine splash during pouring into a gallipot (antiseptic skin preparation container) from heights of 5 cm, 10 cm, 15
Lennie P G Derde et al.
The Lancet. Infectious diseases, 14(1), 31-39 (2013-10-29)
Intensive care units (ICUs) are high-risk areas for transmission of antimicrobial-resistant bacteria, but no controlled study has tested the effect of rapid screening and isolation of carriers on transmission in settings with best-standard precautions. We assessed interventions to reduce colonisation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service