Skip to Content
Merck
All Photos(1)

Key Documents

C46403

Sigma-Aldrich

3-Chloro-1-propanol

98%

Synonym(s):

1-Chloro-3-hydroxypropane, Trimethylene chlorohydrin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl(CH2)3OH
CAS Number:
Molecular Weight:
94.54
Beilstein:
773655
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.445 (lit.)

bp

160-162 °C (lit.)

density

1.131 g/mL at 25 °C (lit.)

SMILES string

OCCCCl

InChI

1S/C3H7ClO/c4-2-1-3-5/h5H,1-3H2

InChI key

LAMUXTNQCICZQX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Improved Synthesis of Pheromone Components: 3-chloro-1-propanol is used as a linchpin to connect the two synthetic blocks in the synthesis of 6,14-dimethyl-1-octadecene. It is likely used as a reactant in a chemical reaction to facilitate the coupling of the two synthetic blocks, enabling the formation of the desired compound (Taguri et al., 2010).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

167.0 °F - closed cup

Flash Point(C)

75 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nico E A Cousens et al.
Faraday discussions, 199, 63-73 (2017-05-06)
Electrowetting-on-dielectric devices typically have operating voltages of 10-20 V. A reduction in the operating voltage could greatly reduce the energy consumption of these devices. Herein, fully reversible one-electrolyte electrowetting of a droplet on a solid metal surface is reported for
A R Jones
Reproduction, fertility, and development, 9(6), 577-581 (1997-01-01)
Boar sperm rapidly interconverted dihydroxyacetone phosphate and glyceraldehyde 3-phosphate, produced fructose-1,6-bisphosphate, approximately equilibrium concentrations of fructose 6-phosphate and glucose 6-phosphate but not glycerol or glycerol 3-phosphate. In the presence of 3-chloro-1-hydroxypropanone, an inhibitor of stage 2 of the glycolytic pathway
A R Jones et al.
Journal of reproduction and fertility, 111(2), 285-290 (1998-02-14)
When incubated in the absence of exogenous substrates, washed boar spermatozoa maintained a high energy charge potential (ECP) for at least 5 h. Addition of 3-chloro-1-hydroxypropanone, an inhibitor of triosephosphate isomerase and glyceraldehyde 3-phosphate dehydrogenase, at any time caused the
Muhammad Ameerullah Sahudin et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 248, 119129-119129 (2020-12-08)
Histamine is one of the important biomarkers for food spoilage in the food sectors. In the present study, a rapid and simple analytical tool has been developed to detect histamine as an indirect strategy to monitor food freshness level. Optical
Hassan M Badawi et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 71(3), 1095-1101 (2008-04-18)
The conformational stability and the three rotor internal rotations in 3-chloro- and 3-bromo-1-propanols were investigated by DFT-B3LYP/6-311+G and ab initio MP2/6-311+G, MP3/6-311+G and MP4(SDTQ)//MP3/6-311+G levels of theory. On the calculated potential energy surface twelve distinct minima were located all of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service